Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SIGLEC H Antibodies

          Invitrogen

          SIGLEC H Monoclonal Antibody (eBio440c), Functional Grade, eBioscience™

          3 Published Figures
          4 References
          View all (11) SIGLEC H antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SIGLEC H Monoclonal Antibody (eBio440c), Functional Grade, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (3)
          SIGLEC H Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          SIGLEC H Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SIGLEC H Antibody (16-0333-82) in Flow

          Cell reports 2020 - Figure 1. Presence of pDCs in Naive Corneas and Alterations in Their Density upon Sterile Inflammation and Acute HSV-1 Keratitis (A and B) Flow cytometric analysis of pooled corneal single-cell suspensions of naive mice (n = 15-20 corneas) and 3 days post-thermal cautery (n = 4-8 corneas). (A) Flow cytometry density plots illustrating PDCA-1 + CD45R/B220 + cells among CD45 + cells in naive and inflamed corn... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SIGLEC H Antibody in Flow Cytometry (Flow)
          SIGLEC H Antibody in Flow Cytometry (Flow)
          SIGLEC H Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          16-0333-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          2 µg/test
          View 4 publications 4 publications

          Functional Assay (Functional)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio440c

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Unconjugated
          • APC 
          • Brilliant UV 615
          • eFluor 450
          • eFluor 660
          • FITC
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10548966

          Product Specific Information

          Description: The monoclonal antibody eBio440c recognizes Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+, and are quick to respond to viruses. Siglec-H is a transmembrane protein of the Ig superfamily that like CD33 have been shown to bind sialic acid but lacks the characteristic cytoplasmic ITIM domain (immunoreceptor tyrosine based inhibitory motif). To overcome the lack of a cytoplasmic domain, Siglec-H associates with DAP12 thereby allowing for signal transduction. The eBio440c antibody has been shown to inhibit pDC function (inhbits IFNalpha secretion in response to CpG).

          It has been observed that some mouse strains (such as SJL) have higher percentages of pDCs compared to C57BL/6.

          Applications Reported: This eBio440c antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBio440c antibody has been tested by blocking staining of the direct conjugate of eBio440c. This can be used at less than or equal to 2.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          It is recommended that direct conjugates such as Product # 11-0333-82 be used rather than purified eBio440c antibody when performing flow cytometric analysis.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. Mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+, and are quick to respond to viruses. Siglec-H is a transmembrane protein of the Ig superfamily that like CD33 have been shown to bind sialic acid but lacks the characteristic cytoplasmic ITIM domain (immunoreceptor tyrosine based inhibitory motif). To overcome the lack of a cytoplasmic domain, Siglec-H associates with DAP12 thereby allowing for signal transduction. The eBio440c antibody has been shown to inhibit pDC function (inhbits IFNalpha secretion in response to CpG). It has been observed that some mouse strains (such as SJL) have higher percentages of pDCs compared to C57BL/6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: mSiglec-H; CD33-related Siglec-like protein; similar to human CD33; sialic acid-binding Ig-like lectins; Siglec; SIGLEC-like protein; unnamed protein product

          View more View less

          Gene Aliases: 6430529G09Rik; Siglec-H

          View more View less

          Entrez Gene ID: (Mouse) 233274

          View more View less

          Function(s)
          protein tyrosine phosphatase activator activity protein phosphatase binding sialic acid binding cargo receptor activity monosaccharide binding
          Process(es)
          immune response-inhibiting signal transduction receptor-mediated endocytosis cell adhesion negative regulation of interleukin-1 beta production negative regulation of interleukin-8 production negative regulation of tumor necrosis factor production Fc-gamma receptor signaling pathway positive regulation of protein secretion negative regulation of inflammatory response cell-cell adhesion negative regulation of monocyte activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-5d76f9f486-n5jxz:80/100.66.73.31:80.
          git-commit: 08b979eaab39376f31a39358ffa32de998e3c7f7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.6.0-offline-prod