Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • Agrin Proteins

          Gibco

          Human Agrin Recombinant Protein, PeproTech®

          View all (2) Agrin proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human Agrin Recombinant Protein, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          160-08-100UG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          ELISA standard (ELISA Std)

          Assay-dependent
          -

          Binding assay (Binding)

          Assay-dependent
          -
          Product Specifications

          Species

          Human

          Expression System

          CHO cells

          Amino acid sequence

          TCPERALERR EEEANVVLTG TVEEILNVDP VQHTYSCKVR VWRYLKGKDL VARESLLDGG NKVVISGFGD PLICDNQVST GDTRIFFVNP APPYLWPAHK NELMLNSSLM RITLRNLEEV EFCVEDKPGT HFTPVPPTPP DACRGMLCGF GAVCEPNAEG PGRASCVCKK SPCPSVVAPV CGSDASTYSN ECELQRAQCS QQRRIRLLSR GPCGSRDPCS NVTCSFGSTC ARSADGLTAS CLCPATCRGA PEGTVCGSDG ADYPGECQLL RRACARQENV FKKFDGPCDP CQGALPDPSR SCRVNPRTRR PEMLLRPESC PARQAPVCGD DGVTYENDCV MGRSGAARGL LLQKVRSGQC QGRDQCPEPC RFNAVCLSRR GRPRCSCDRV TCDGAYRPVC AQDGRTYDSD CWRQQAECRQ QRAIPSKHQG PCDQAPSPCL GVQCAFGATC AVKNGQAACE CLQACSSLYD PVCGSDGVTY GSACELEATA CTLGREIQVA RKGPCDRCGQ CRFGALCEAE TGRCVCPSEC VALAQPVCGS DGHTYPSECM LHVHACTHQI SLHVASAGPC ETCGDAVCAF GAVCSAGQCV CPRCEHPPPG PVCGSDGVTY GSACELREAA CLQQTQIEEA RAGPCEQAEC GSGGSGSGED GDCEQELCRQ RGGIWDEDSE DGPCVCDFSC QSVPGSPVCG SDGVTYSTEC ELKKARCESQ RGLYVAAQGA CRGPTFAPLP PVAPLHCAQT PYGCCQDNIT AARGVGLAGC PSACQCNPHG SYGGTCDPAT GQCSCRPGVG GLRCDRCEPG FWNFRGIVTD GRSGCTPCSC DPQGAVRDDC EQMTGLCSCK PGVAGPKCGQ CPDGRALGPA GCEADASAPA TCAEMRCEFG ARCVEESGSA HCVCPMLTCP EANATKVCGS DGVTYGNECQ LKTIACRQGL QISIQSLGPC QEAVAPSTHP TSASVTVTTP GLLLSQALPA PPGALPLAPS STAHSQTTPP PSSRPRTTAS VPRTTVWPVL TVPPTAPSPA PSLVASAFGE SGSTDGSSDE ELSGDQEASG GGSGGLEPLE GSSVATPGPP VER

          Molecular weight

          111.9 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 95% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by its ability to bind U-87 MG cells in a functional ELISA.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          100 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          Recombinant Human Agrin is 111.9 kDa protein containing 1073 amino acids, corresponding to the 110 kDa N-terminal subunit of the full length protein. As a result of glycosylation, Recombinant Human Agrin migrates with an apparent molecular mass of approximately 150-160 kDa by SDS-PAGE gel, under reducing and non-reducing conditions.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          Agrin is a molecule that resides in the basal lamina of muscle cells and directs key events in post synaptic differentiation. Most notably, Agrin is responsible for the clustering of acetylcholine receptors (AChRs) on the cell surface and their localization to the neuromuscular junction. Several Agrin variants have been identified which arise from alternative mRNA splicings. Agrin splice forms having inserts at two sites in the carboxy terminus designated "y" and "z" display a high affinity for AChRs, while splice forms lacking these inserts associate with AChRs weakly. Muscle alpha-dystroglycan has been postulated to be the receptor for the clustering activity of agrin; however, this is a point of contention. Tyrosine phosphorylation has been implicated as a required early step in AChR aggregation. Interestingly, a unique receptor tyrosine kinase, designated MuSK, has been discovered that interacts with Agrin and is specifically localized to developing muscle.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: agrin proteoglycan; RP23-139J18.2; synaptic basal lamina component; unnamed protein product

          View more View less

          Gene Aliases: AGRIN; CMS8; CMSPPD

          View more View less

          UniProt ID: (Human) O00468

          View more View less

          Entrez Gene ID: (Human) 375790

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0