2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT
2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT
2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT
Thermo Scientific Chemicals

2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT

CAS: 513-81-5 | C6H10 | 82.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 163430250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS513-81-5
SpecificationsSpecification SheetSpecification Sheet
Stabilizer100 ppm BHT
Appearance (Color)Clear colorless
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.4370 to 1.4400 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only