Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ATP1A2 Antibodies

          Proteintech

          ATP1A2 Polyclonal Antibody

          20 References
          View all (8) ATP1A2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ATP1A2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (21)
          ATP1A2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ATP1A2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ATP1A2 Antibody (16836-1-AP) in ICC/IF

          Immunofluorescent analysis of Hela cells, using ATP1A2 antibody 16836-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ATP1A2 Antibody in Immunocytochemistry (ICC/IF)
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ATP1A2 Antibody in Western Blot (WB)
          ATP1A2 Antibody in Western Blot (WB)
          ATP1A2 Antibody in Western Blot (WB)
          ATP1A2 Antibody in Western Blot (WB)
          ATP1A2 Antibody in Flow Cytometry (Flow)
          ATP1A2 Polyclonal Antibody

          Product Details

          16836-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          View 12 publications 12 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          0.5 µg/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Dog, Human, Mollusc, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ATP1A2 Fusion Protein Ag10515 (347-700 aa encoded by BC052271)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          31.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody was raised against the internal region of the human ATP1A2 and can recognize all the isoforms of alpha subunit. The 65 kDa band detected occasionally may be the degradation product of ATP1A2.

          Immunogen sequence: KRMARKNCL VKNLEAVETL GSTSTICSDK TGTLTQNRMT VAHMWFDNQI HEADTTEDQS GATFDKRSPT WTALSRIAGL CNRAVFKAGQ ENISVSKRDT AGDASESALL KCIELSCGSV RKMRDRNPKV AEIPFNSTNK YQLSIHERED SPQSHVLVMK GAPERILDRC STILVQGKEI PLDKEMQDAF QNAYMELGGL GERVLGFCQL NLPSGKFPRG FKFDTDELNF PTEKLCFVGL MSMIDPPRAA VPDAVGKCRS AGIKVIMVTG DHPITAKAIA KGVGIISEGN ETVEDIAARL NIPMSQVNPR EAKACVVHGS DLKDMTSEQL DEILKNHTEI VFARTSPQQK LIIVE (347-700 aa encoded by BC052271)

          Target Information

          Atp1a2 encodes an Na+/K+ ATPase, which is a membrane protein that is composed of two subunits - alpha and beta. The Na+/K+ ATPase pump maintains an essential electrochemical gradient within cells through the active transport of sodium (Na+) and potassium (K+) ions. The Atp1a2 encoded protein belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These ion gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. The ATPase enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta), and the catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. The Atp1a2 gene encodes an alpha 2 subunit. Mutations in the Atp1a2 gene result in familial basilar or hemiplegic migraines, and in a rare syndrome known as alternating hemiplegia of childhood. Diseases associated with ATP1A2 include Alternating Hemiplegia Of Childhood and Migraine, Familial Hemiplegic, 2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase, Na+K+ transporting, alpha 2; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ -ATPase alpha 2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; Na,K-ATPase subunit alpha-B; Sodium Potassium ATPase; Sodium pump subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; Sodium/potassium-transporting ATPase subunit alpha-2; unnamed protein product

          View more View less

          Gene Aliases: ATP1A2; Atpa-3; AW060654; DEE98; FARIMPD; FHM2; KIAA0778; MHP2; mKIAA0778; RATATPA2

          View more View less

          UniProt ID: (Human) P50993, (Rat) P06686, (Mouse) Q6PIE5

          View more View less

          Entrez Gene ID: (Human) 477, (Rat) 24212, (Mouse) 98660

          View more View less

          Function(s)
          nucleotide binding sodium:potassium-exchanging ATPase activity steroid binding protein binding ATP binding potassium-transporting ATPase activity phosphatase activity ATPase activity cation-transporting ATPase activity potassium ion binding sodium ion binding metal ion binding protein heterodimerization activity chaperone binding steroid hormone binding hydrolase activity primary active transporter
          Process(es)
          neurotransmitter uptake behavioral fear response regulation of the force of heart contraction regulation of respiratory gaseous exchange by neurological system process ion transport potassium ion transport sodium ion transport cellular sodium ion homeostasis regulation of muscle contraction regulation of smooth muscle contraction regulation of striated muscle contraction cell communication blood circulation regulation of heart contraction regulation of blood pressure adult locomotory behavior visual learning establishment or maintenance of transmembrane electrochemical gradient regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion response to auditory stimulus neuronal action potential propagation regulation of vasoconstriction L-ascorbic acid metabolic process amygdala development olfactory cortex development cellular cation homeostasis cellular potassium ion homeostasis response to nicotine locomotory exploration behavior sodium ion transmembrane transport response to potassium ion sodium ion export from cell locomotion fear response negative regulation of heart contraction positive regulation of heart contraction negative regulation of striated muscle contraction ATP metabolic process negative regulation of cytosolic calcium ion concentration membrane depolarization regulation of glutamate uptake involved in transmission of nerve impulse regulation of synaptic transmission, glutamatergic relaxation of cardiac muscle cardiac muscle contraction cellular response to mechanical stimulus cellular response to steroid hormone stimulus potassium ion transmembrane transport regulation of cardiac muscle cell contraction membrane repolarization membrane depolarization during cardiac muscle cell action potential cell communication by electrical coupling involved in cardiac conduction cation transmembrane transport transport across blood-brain barrier hydrogen ion transmembrane transport negative regulation of calcium ion transmembrane transport response to glycoside potassium ion import across plasma membrane metabolic process potassium ion import regulation of cardiac muscle contraction by calcium ion signaling energy coupled proton transmembrane transport, against electrochemical gradient ATP hydrolysis coupled proton transport negative regulation of calcium:sodium antiporter activity transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline