Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD90.1 (Thy1.1) Antibodies

          Invitrogen

          CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), APC, eBioscience™

          45 Published Figures
          90 References
          View all (18) CD90.1 (Thy1.1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), APC, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (45)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 46

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD90.1 (Thy-1.1) Antibody (17-0900-82) in Flow

          Staining of rat splenocytes with Anti-Rat/Mouse CD90.1 FITC (left), and PE (right). Appropriate isotype controls were used (open histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          17-0900-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.06 µg/test
          View 88 publications 88 publications

          Affinity Purification (AP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mammal, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a, kappa

          Recommended Isotype Control

          Mouse IgG2a kappa Isotype Control (eBM2a), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HIS51

          Conjugate

          APC APC APC
          • Unconjugated
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 615
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_469420

          Product Specific Information

          Description: The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells.

          Applications Reported: The HIS51 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The HIS51 antibody has been tested by flow cytometric analysis of rat thymocyte and splenocyte suspensions. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD90; cell surface protein; FLJ33325; thy-1 antigen; thy-1 glycoprotein; Thy-1 protein; Thy-1 protein precursor; thymus cell antigen 1, theta; Thymus cell surface antigen; unnamed protein product

          View more View less

          Gene Aliases: CD7; CD90; T25; Thy-1; Thy-1.2; Thy1.1; Thy1.2

          View more View less

          UniProt ID: (Rat) P01830, (Mouse) P01831

          View more View less

          Entrez Gene ID: (Rat) 24832, (Mouse) 21838

          View more View less

          Function(s)
          GTPase activator activity integrin binding protein binding enzyme binding protein kinase binding GPI anchor binding
          Process(es)
          angiogenesis regulation of cell-matrix adhesion regulation of immune system process positive regulation of cellular extravasation negative regulation of protein kinase activity cytoskeleton organization cell adhesion cell surface receptor signaling pathway integrin-mediated signaling pathway cell-cell signaling regulation of cell migration negative regulation of cell migration heterotypic cell-cell adhesion negative regulation of apoptotic process receptor clustering positive regulation of GTPase activity mast cell activation positive regulation of cell adhesion retinal cone cell development protein autophosphorylation focal adhesion assembly negative regulation of fibroblast proliferation response to axon injury positive regulation of peptidyl-tyrosine phosphorylation negative regulation of axonogenesis T cell receptor signaling pathway negative regulation of T cell receptor signaling pathway positive regulation of T cell activation positive regulation of release of sequestered calcium ion into cytosol positive regulation of focal adhesion assembly negative regulation of neuron projection regeneration cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-ncdd4:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline