Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • MHC Class II (I-A/I-E) Antibodies

          Invitrogen

          MHC Class II (I-A/I-E) Monoclonal Antibody (M5/114.15.2), APC, eBioscience™

          82 Published Figures
          169 References
          View all (54) MHC Class II (I-A/I-E) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MHC Class II (I-A/I-E) Monoclonal Antibody (M5/114.15.2), APC, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (82)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 83

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MHC Class II (I-A/I-E) Antibody (17-5321-82) in Flow

          Staining of C57BL/6 splenocytes with Anti-Mouse CD3e FITC (Product # 11-0031-82) and 0.015 µg of Rat IgG2b K Isotype Control APC (Product # 17-4031-82) (left) or 0.015 µg of Anti-Mouse MHC Class II (I-A/I-E) APC (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)
          MHC Class II (I-A/I-E) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          17-5321-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.03 µg/test
          View 163 publications 163 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Bacteria, Human, Mouse, Rat

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          M5/114.15.2

          Conjugate

          APC APC APC
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 700 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 496
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant Violet 421
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 610-70S
          • NovaFluor Blue 660-120S
          • NovaFluor Blue 725
          • NovaFluor Blue 760
          • NovaFluor Blue 800
          • NovaFluor Red 685
          • NovaFluor Red 710
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Violet 745
          • NovaFluor Violet 800
          • NovaFluor Yellow 570
          • NovaFluor Yellow 590
          • NovaFluor Yellow 660
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_469455

          Product Specific Information

          Description: The M5/114.15.2 monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.

          Applications Reported: M5/114.15.2 has been reported for use in flow cytometric analysis.

          Applications Tested: The M5/114.15.2 antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 0.03 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.

          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cell surface glycoprotein; class II Ia antigen A-beta; H-2 class II histocompatibility antigen, A beta chain; H-2 class II histocompatibility antigen, A-B alpha chain; H-2 class II histocompatibility antigen, A-F alpha chain; H-2 class II histocompatibility antigen, A-K alpha chain; H-2 class II histocompatibility antigen, A-Q alpha chain; H-2 class II histocompatibility antigen, A-R alpha chain; H-2 class II histocompatibility antigen, A-S alpha chain; H-2 class II histocompatibility antigen, A-U alpha chain; H2-I-E-beta; H2-IA-beta cell surface glycoprotein; histocompatibility 2, class II antigen A, alpha, promoter 1; histocompatibility 2, class II antigen A, beta, promoter 1; histocompatibility 2, class II antigen E beta, promoter 1; I-Ab; I-Ad; I-Aq; I-Ar MHC class II molecule alpha chain; I-Ar MHC class II molecule beta chain; I-E beta MHC class II; I-Ed; I-Ek; IA-alpha polyprotein; major histocompatibility complex class II beta chain; major histocompatibility complex H-2E beta chain; major histocompatibility protein class II alpha chain; MHC class 2; MHC class II antigen alpha chain; MHC class II H2-IA-beta-psi; MHC class II IE-b; MHC E-beta-b precursor; MHC H2-I-A-beta chain; MHC H2-IA-alpha; MHC H2-IE-beta; MHC H2-IE-beta cell surface glycoprotein; MHC NOD beta chain precursor; MHC-II I-A(d) alpha; MHC-II I-A(d) beta; response to metastatic cancers 1; unnamed protein product

          View more View less

          Gene Aliases: Aalpha; Abeta; Eb; H-2Aa; H-2Ab; H-2Eb; H2-Ab; H2Aa; H2Eb; I-Aalpha; I-Abeta; Ia-1; Ia-2; Ia-4; Ia1; Ia2; Ia4; IAalpha; IAb; Rmcs1

          View more View less

          UniProt ID: (Mouse) P14434, (Mouse) P14483

          View more View less

          Entrez Gene ID: (Mouse) 14960, (Mouse) 14961, (Mouse) 14969

          View more View less

          Function(s)
          protein binding MHC class II protein complex binding peptide antigen binding macromolecular complex binding polysaccharide binding T cell receptor binding CD4 receptor binding
          Process(es)
          immune system process peptide antigen assembly with MHC class II protein complex immune response antigen processing and presentation antigen processing and presentation of exogenous peptide antigen via MHC class II positive regulation of immune response positive regulation of T cell activation negative regulation of T cell proliferation positive regulation of T cell differentiation antigen processing and presentation of peptide antigen
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-6qggw:80/100.66.76.149:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline