Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FLK1 (VEGF Receptor 2) Antibodies

          Invitrogen

          CD309 (FLK1) Monoclonal Antibody (Avas12a1), APC, eBioscience™

          Advanced Verification
          46 Published Figures
          44 References
          View all (97) FLK1 (VEGF Receptor 2) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD309 (FLK1) Monoclonal Antibody (Avas12a1), APC, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (46)
          • Advanced Verification (1)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 48

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD309 (FLK1) Antibody (17-5821-80) in Flow

          Staining of the bEnd-3 cell line with 0.25 µg of Rat IgG2a K Isotype Control APC (Product # 17-4321-81) (open histogram) or 0.25 µg of Anti-Mouse CD309 (FLK1) APC (filled histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody in Flow Cytometry (Flow)
          CD309 (FLK1) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          17-5821-80

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.5 µg/test
          View 42 publications 42 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Avas12a1

          Conjugate

          APC APC APC
          • Unconjugated
          • Alexa Fluor 700 
          • Biotin 
          • PE 
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_657865

          Product Specific Information

          Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.

          Applications Reported: This Avas12a1 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This Avas12a1 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD309 antigen; fetal liver kinase 1; Flk-1; kinase NYK; platelet-derived growth factor receptor-related tyrosine kinase; protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; unnamed protein product; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor; VEGF receptor-2

          View more View less

          Gene Aliases: 6130401C07; Flk-1; Flk1; Krd-1; Ly73; orv; sVEGFR-2; VEGFR-2; VEGFR2

          View more View less

          UniProt ID: (Mouse) P35918

          View more View less

          Entrez Gene ID: (Mouse) 16542

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity vascular endothelial growth factor-activated receptor activity integrin binding protein binding ATP binding coreceptor activity growth factor binding vascular endothelial growth factor binding identical protein binding cadherin binding
          Process(es)
          angiogenesis ovarian follicle development patterning of blood vessels vasculogenesis response to hypoxia positive regulation of protein phosphorylation regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation lymph vessel development positive regulation of mesenchymal cell proliferation negative regulation of systemic arterial blood pressure endocardium development endothelium development endochondral bone growth transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of cytosolic calcium ion concentration positive regulation of cell proliferation regulation of cell shape gonad development male gonad development response to xenobiotic stimulus positive regulation of signal transduction positive regulation of endothelial cell migration negative regulation of gene expression positive regulation of macroautophagy cell migration peptidyl-tyrosine phosphorylation hemopoiesis lung development positive regulation of cell migration positive regulation of BMP signaling pathway positive regulation of vascular endothelial growth factor receptor signaling pathway positive regulation of TOR signaling embryonic hemopoiesis cellular response to vascular endothelial growth factor stimulus vascular endothelial growth factor receptor-2 signaling pathway vascular endothelial growth factor signaling pathway vasodilation surfactant homeostasis regulation of MAPK cascade positive regulation of MAPK cascade negative regulation of neuron apoptotic process positive regulation of blood vessel endothelial cell migration cell fate commitment endothelial cell differentiation positive regulation of angiogenesis protein autophosphorylation vascular endothelial growth factor receptor signaling pathway positive regulation of long-term neuronal synaptic plasticity lung alveolus development cell maturation blood vessel morphogenesis post-embryonic camera-type eye morphogenesis branching morphogenesis of an epithelial tube neuron projection morphogenesis positive regulation of epithelial cell proliferation positive regulation of calcium-mediated signaling positive regulation of positive chemotaxis positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling positive regulation of mitochondrial depolarization calcium ion homeostasis epithelial tube morphogenesis blood vessel endothelial cell differentiation vascular wound healing endothelial tube morphogenesis positive regulation of ERK1 and ERK2 cascade semaphorin-plexin signaling pathway positive regulation of cell migration involved in sprouting angiogenesis positive regulation of mitochondrial fission positive regulation of nitric oxide-cGMP mediated signal transduction regulation of hematopoietic progenitor cell differentiation regulation of bone development cellular response to hydrogen sulfide negative regulation of endothelial cell apoptotic process positive regulation of endothelial cell chemotaxis positive regulation of vasculogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline