TEMED - FAQs
TEMED - FAQs
View additional product information for TEMED - FAQs (17919)
5 product FAQs found
What is the recommended storage condition for TEMED?
We recommend storing TEMED locked up in a segregated and approved area. Store in the original container protected from direct sunlight in a dry, cool and well-ventilated area and separate from oxidizing materials.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What are the Quality Specifications for each lot of TEMED?
The Certificate of Analysis lists actual test results for each lot and the minimum specification requirements:
-Purity > 99.0%
-Refractive index (nD20) between 1.417-1.419
-Boiling Range: 119-121 degrees C
-The solution must be a clear liquid, free of particulate matter
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What are the characteristics of TEMED (Cat. No. 17919)?
>Chemical name: N,N,N',N'- tetramethylethane-1,2-diamine
-Formula: C6H16N2
-CAS number: 110-18-9
-Molecular weight: 116.21-Purity: ≥ 99%
-Refractive index: 1.417 to 1.419
-Boiling range: 119 to 121 degrees C
-Clear liquid at room temperature, sold as a 25 mL volume in an amber bottle
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is Thermo Scientific Tetramethylethylenediamine (TEMED)?
Thermo Scientific Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is used with ammonium persulfate (APS) to catalyze acrylamide polymerization when preparing gels for electrophoresis.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.