TEMED
Thermo Scientific™

TEMED

Thermo Scientific Pierce Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is used with ammonium persulfate (APS)Read more
Have Questions?
Catalog NumberQuantity
1791925 mL
Catalog number 17919
Price (USD)
91.75
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
91.75
Each
Thermo Scientific Pierce Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is used with ammonium persulfate (APS) to catalyze acrylamide polymerization when preparing gels for electrophoresis.

Features of Thermo Scientific Pierce Tetramethylethylenediamine (TEMED) :

• Chemical name: N,N,N',N'-tetramethylethane-1,2-diamine
• Formula: C6H16N2
• CAS number: 110-18-9
• Molecular weight: 116.21
• Purity: ≥99%
• Refractive index: 1.417 to 1.419
• Boiling range: 119 to 121°C
• Clear liquid at room temperature
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Quantity25 mL
Product TypeTEMED
Unit SizeEach
Contents & Storage
Store locked up in a segregated and approved area. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, and separate from oxidizing materials.

Frequently asked questions (FAQs)

What is the recommended storage condition for TEMED?

We recommend storing TEMED locked up in a segregated and approved area. Store in the original container protected from direct sunlight in a dry, cool and well-ventilated area and separate from oxidizing materials.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the Quality Specifications for each lot of TEMED?

The Certificate of Analysis lists actual test results for each lot and the minimum specification requirements:
-Purity > 99.0%
-Refractive index (nD20) between 1.417-1.419
-Boiling Range: 119-121 degrees C
-The solution must be a clear liquid, free of particulate matter

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the characteristics of TEMED (Cat. No. 17919)?

>Chemical name: N,N,N',N'- tetramethylethane-1,2-diamine
-Formula: C6H16N2
-CAS number: 110-18-9
-Molecular weight: 116.21-Purity: ≥ 99%
-Refractive index: 1.417 to 1.419
-Boiling range: 119 to 121 degrees C
-Clear liquid at room temperature, sold as a 25 mL volume in an amber bottle


Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is Thermo Scientific Tetramethylethylenediamine (TEMED)?

Thermo Scientific Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is used with ammonium persulfate (APS) to catalyze acrylamide polymerization when preparing gels for electrophoresis.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do you carry ammonium persulfate and TEMED?

Yes, details below:

- Ammonium Persulfate: Cat. No. 17874
- TEMED: Cat. No. 15524010 and Cat. No. 17919

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.