Is it normal to see a band in the SuperScript IV First-Strand Synthesis System (Cat. Nos. 18091050, 18091200) "No Reverse Transcriptase (RT) Control"?
Yes. The SuperScript IV First-Strand Synthesis System (Cat. No. 18091050, 18091200) "No Reverse Transcriptase (RT) Control" will often display a level of amplification because of residual genomic DNA in the RNA preps. In general, you do not need to worry if a faint band shows up on a gel, as long as the intensity of the positive control is much higher.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.