SuperScript™ IV First-Strand Synthesis System
Invitrogen™

SuperScript™ IV First-Strand Synthesis System

The SuperScript IV First-Strand Synthesis System for RT-PCR, available with or without ezDNase, is optimized for synthesis of first-strand cDNA from purified poly(A)+ or total RNA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesNo. of Reactions
18091050Kit only50 Reactions
18091200Kit only200 Reactions
18091150Kit with ezDNase50 Reactions
18091300Kit with ezDNase200 Reactions
Catalog number 18091050
Price (KRW)
687,000
Online offer
Ends: 31-Dec-2026
808,000
Save 121,000 (15%)
Each
Includes:
Kit only
No. of Reactions:
50 Reactions
Request bulk or custom format
Price (KRW)
687,000
Online offer
Ends: 31-Dec-2026
808,000
Save 121,000 (15%)
Each
The SuperScript IV First-Strand Synthesis System for RT-PCR, available with or without ezDNase, is optimized for synthesis of first-strand cDNA from purified poly(A)+ or total RNA. The system includes SuperScript IV Reverse Transcriptase (RT), a proprietary MMLV mutant that produces robust and reliable cDNA synthesis in RT reactions. The SuperScript IV First-Strand Synthesis System with ezDNase has the further ability to remove potential genomic DNA contamination.

The SuperScript IV synthesis system is significantly improved over the SuperScript III-based system in inhibitor resistance, processivity, and reaction speed, while retaining all the benefits of the previous system, including increased thermostability, highly efficient full-length cDNA synthesis, and reduced RNase activity. The SuperScript IV system is designed to provide reliable, consistent, and fast cDNA synthesis in the presence of inhibitors found in a wide variety of samples that cause other currently available RTs to perform inefficiently.

The extremely simplified genomic DNA removal step of the SuperScript IV First-Strand Synthesis System with ezDNase dramatically reduces the time of the entire reverse transcription protocol and reduces possible variation in gene expression due to RNA loss or damage during conventional DNase treatment. The SuperScript IV synthesis system with ezDNase is the top choice for performance and flexibility for RT-PCR applications. We recommend it for all RT-PCR applications, especially when reproducibility and reliability is the primary concern and when inhibitors in the RNA sample may interfere with cDNA synthesis, leading to biases in gene expression studies.

Both SuperScript IV First-Strand Synthesis systems contain all components needed for RT reactions, plus an additional control gene and primers, and provide the flexibility to customize the RT set-up to fit the needs of your application.

Features of the SuperScript IV First-Strand Synthesis System include:
• Significantly improved resistance to a variety of inhibitors that can interfere with cDNA synthesis
• Robust and specific cDNA synthesis in a wide range of sample types
• A faster reverse transcriptase reaction that reduces incubation time from >50 min to 10 min
• Significantly better processivity compared to SuperScript III RT
• Simplified genomic DNA removal step (SuperScript IV synthesis system with ezDNase only)

Enzyme source
Purified from E. coli expressing the pol gene of M-MLV, modified to increase thermostability, half-life, processivity, and inhibitor resistance and to reduce RNase H activity.

Performance and quality testing
Endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease assays; and yield and length of cDNA product

Unit definition
One unit of SuperScript IV RT is the amount of enzyme required to incorporate 1 nmole of deoxyribonucleotide into acid-precipitable material in 10 min at 37°C using poly(A) oligo(dT)12-18 as a template/primer.

Unit reaction conditions
50 mM Tris-HCl (pH 8.3), 4 mM MgCl2, 10 mM DTT, 50 mM KCl, 0.5 mM dTTP, 0.4 MBq/mL [3H]-dTTP, 0.4 mM poly(A) oligo (dT)12-18 and enzyme in 20 μL for 10 min at 37°C

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
IncludesKit only
No. of Reactions50 Reactions
Optimal Reaction Temperature50°C
QuantityEach
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseSuperScript IV
Shipping ConditionDry Ice
Size (Final Product)Up to 12.3 kb
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)RT-PCR, Real Time PCR (qPCR)
GC-Rich PCR PerformanceHigh
Reaction Speed10 min.
Unit SizeEach
Contents & Storage

• Oligo(dT)20 , 50 μL (50 μM)
• Random hexamers, 250 μL (50 ng/μL)
• 5X RT buffer, 1 mL
• DTT, 250 μL (0.1 M)
• dNTP mix, 250 μL (10 mM)
• SuperScript IV RT, 50 μL (200 U/μL)
• Ribonuclease Inhibitor, 100 μL (40 U/μL)
• E. coli RNase H, 50 μL (2 U/μL)
• DEPC-treated water, 1.2 mL
• Total HeLa RNA, 20 μL (10 ng/μL)
• Sense Control Primer, 25 μL (10 μM)
• Antisense Control Primer, 25 μL (10 μM)

Store at -15°C to -25°C.

Frequently asked questions (FAQs)

Is it normal to see a band in the SuperScript IV First-Strand Synthesis System (Cat. Nos. 18091050, 18091200) "No Reverse Transcriptase (RT) Control"?

Yes. The SuperScript IV First-Strand Synthesis System (Cat. No. 18091050, 18091200) "No Reverse Transcriptase (RT) Control" will often display a level of amplification because of residual genomic DNA in the RNA preps. In general, you do not need to worry if a faint band shows up on a gel, as long as the intensity of the positive control is much higher.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

In your SuperScript IV RT protocols, there is no ezDNase inactivation step. Will ezDNase be active to affect RNA or the RT reaction?

The Invitrogen ezDNase Enzyme is a novel DNase that is highly specific for double-stranded DNA. It has no activity on single-stranded DNA in RT reactions (primers or probes), or on RNA. The enzyme is also thermolabile—it is inactivated quickly at temperatures typical for the SuperScript IV RT reaction (e.g., 50 degrees C). The additional inactivation step is therefore not required for most RT-qPCR applications. If the RNA sample is to be used for RT-PCR of fragments ≥3 kb, incubate the sample for 5 minutes at 55 degrees C in the presence of 10 mM DTT to inactivate the enzyme.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Which SuperScript IV RT format do you recommend for real-time PCR applications?

For RT-qPCR applications we recommend using the Invitrogen SuperScript IV VILO Master Mix. The cDNA synthesis reaction setup with this master mix requires fewer pipetting steps and therefore reduces variation in the data. SuperScript IV RT, as a component of the master mix, offers the highest efficiency of cDNA synthesis step compared to competitors’ products.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Are there any significant changes in the SuperScript IV RT protocol compared to the SuperScript III RT protocol?

The only difference is that the incubation time for the reverse transcription reaction has been reduced to 10 minutes and inactivation time has been reduced to 5 minutes at 85 degrees C.

Can I get comparable cDNA yield and length using the SuperScript IV RT 10-minute protocol as when using the 50-minute protocol for SuperScript III RT?

When compared with SuperScript III RT (and other manufacturers’ RTs) in a synthesis reaction for a 9 kb cDNA, SuperScript IV RT performed successful synthesis in just 10 minutes and did so with comparable (or improved) yield (as shown by gel band density).

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.