Can exo-Klenow be purchased separately from your array labeling kits?
Yes, the catalog number for the exo-Klenow is AM2008. The enzyme concentration is 5 U/µL.
Search
View additional product information for BioPrime™ Array CGH Genomic Labeling Module - FAQs (18095012)
3 product FAQs found