Phloxine B, 85%, pure, high purity, Biological stain
Phloxine B, 85%, pure, high purity, Biological stain
Phloxine B, 85%, pure, high purity, Biological stain
Thermo Scientific Chemicals

Phloxine B, 85%, pure, high purity, Biological stain

CAS: 18472-87-2 | C20H2Br4Cl4Na2O5
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 189470050
Price (USD)
61.30
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
61.30
Each
Chemical Identifiers
CAS18472-87-2
IUPAC Namedisodium 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular FormulaC20H2Br4Cl4Na2O5
InChI KeyOOYIOIOOWUGAHD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C1=C(Br)C=C2C(OC3=C(Br)C([O-])=C(Br)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<10 % (105°C)
UV Lambda max546 to 550 nm (in 50:50 METHS:water)
Specific absorbance A (1%/1cm)875 to 1300 (at lambda max.)
Infrared spectrumConforms
Absorption Ratio P-15/P+151.38 to 2.24
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only