Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Proteins & Peptides ›
          • IL18BP Proteins

          Gibco

          Human IL-18BP Fc Recombinant Protein, PeproTech®

          2 References
          View all (2) IL18BP proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human IL-18BP Fc Recombinant Protein, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          200-18BP-1MG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 2 publications 2 publications

          Bioactivity (Bioactivity)

          Assay-dependent
          -
          Product Specifications

          Species

          Human

          Published species

          Human

          Expression System

          CHO cells

          Amino acid sequence

          TPVSQTTTAA TASVRSTKDP CPSQPPVFPA AKQCPALEVT WPEVEVPLNG TLSLSCVACS RFPNFSILYW LGNGSFIEHL PGRLWEGSTS RERGSTGTQL CKALVLEQLT PALHSTNFSC VLVDPEQVVQ RHVVLAQLWA GLRATLPPTQ EALPSSHSSP QQQGGPKSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK

          Tag

          Fc-tag

          Molecular weight

          87.2 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 95% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by its ability to neutralize IL-18 induced IFN-gamma production in KG-1 cells.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          1 mg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          PeproTech's CHO cell-derived Recombinant Human IL-18BP Fc is a glycosylated, disulfide-linked homodimer of 792 amino-acid-residues whose monomer consists of a 163-amino-acid portion of secreted IL-18BP fused to the 231-amino-acid length Fc portion of human IgG by two glycine residues. The calculated molecular weight of monomeric CHO cell-derived Recombinant Human IL-18BP Fc is 43.6 kDa; however, due to glycosylation, it migrates at an apparent molecular weight of approximately 60-65 kDa by SDS-PAGE analysis under reducing conditions.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          Interleukin-18 binding protein (IL-18BP) is a constitutively secreted glycoprotein that acts as a natural antagonist for interleukin-18 (IL-18) by preventing interaction between IL-18 and its receptors. Through this binding, IL-18BP neutralizes the proinflammatory role of IL-18 in cell-mediated immune responses, resulting in reduced IFN-gamma production by T helper type I cells and inhibition of allospecific cytotoxic T lymphocyte (CTL) activity through augmented natural killer (NK) cell cytotoxicity. As an inhibitor of IL-18 and an essential component of skin homeostasis, IL-18BP has been linked to a variety of diseases, including eczema, asthma, and the autoimmune condition, Crohn's disease. The upregulation of IL-18BP by keratinocytes expressing the human papillomavirus (HPV) E7 oncoprotein has been shown to decrease IL-18-induced IFN-gamma production and IL-18-mediated T cell activation. Elevated levels of IL-18 and IL-18BP, as well as proinflammatory cytokines IFN-gamma, TNF-alpha, IL-1beta and IL-8, have been linked to chronic inflammation and observed in macrophages (including Kupffer cells) isolated from lesions and intestinal tissues of Crohn's disease patients. As a member of the immunoglobulin-like class of receptors, IL-18BP contains a single immunoglobulin (Ig) domain; although two (b and d) of the four identified human isoforms (a-d) contain incomplete domains that greatly reduce their binding affinity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: IL-18BP; IL-18BPa; IL-18BPb; IL-18BPc; Interleukin-18-binding protein; MC51L-53L-54L homolog gene product; Tadekinig-alfa; unnamed protein product

          View more View less

          Gene Aliases: FVH; IL18BP; IL18BPa

          View more View less

          UniProt ID: (Human) O95998

          View more View less

          Entrez Gene ID: (Human) 10068

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline