Pierce™ Avidin Agarose
Thermo Scientific™

Pierce™ Avidin Agarose

Thermo Scientific Pierce Avidin Agarose can be used in a variety of applications for the affinity purification of biotinylated macromolecules.Features자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
202195 mL
202255 x 5 mL
카탈로그 번호 20219
제품 가격(KRW)
376,000
온라인 행사
Ends: 30-Sep-2026
442,000
할인액 66,000 (15%)
Each
수량:
5 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
376,000
온라인 행사
Ends: 30-Sep-2026
442,000
할인액 66,000 (15%)
Each
Thermo Scientific Pierce Avidin Agarose can be used in a variety of applications for the affinity purification of biotinylated macromolecules.

Features of Avidin Agarose:

• Strong , nearly irreversible biotin:avidin interaction
• Binding biotinylated anti-transferrinfrom serum (1)
• Binding biotinyl peptides and elution with a SDS/Urea solution (2)
• Hybridization of biotinylated RNA to its complementary DNA and binding to immobilized avidin, with subsequent elution of the single-stranded DNA (3)
• Purification of double-stranded DNA (4)
• Approximate Binding Capacity >20 μg biotin/mL resin

In one variation, an antibody that has an affinity for a particular antigen is labeled with biotin. Cells containing the antigen are lysed, then incubated with the biotinylated antibody to form a typical antigen/antibody complex. To isolate the antigen, the crude mixture is passed through an immobilized avidin or streptavidin column, which will bind the complex. After appropriate washes, the antigen can then be eluted from the column with a pH 2.8 elution buffer. The biotinylated antibody is retained by the column.

Related Products
Pierce™ Monomeric Avidin Agarose
For Research Use Only. Not for use in diagnostic procedures.
사양
컬럼 유형Affinity, Agarose Resin
형식Bottle
수량5 mL
정지상Avidin
타겟Biotin-Tagged Molecule
용량(미터법)20 μg Biotin Binding per mL Resin
제품라인Pierce
유형Agarose
Unit SizeEach
구성 및 보관
Store at 4°C. Do not freeze.