Pierce™ Streptavidin Agarose Columns, 1 mL - FAQs
Pierce™ Streptavidin Agarose Columns, 1 mL - FAQs
View additional product information for Pierce™ Streptavidin Agarose Columns, 1 mL - FAQs (20351)
1 product FAQs found
How many biotin binding-sites are there per streptavidin molecule?
Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.