Pierce™ Streptavidin Agarose Columns, 1 mL
Thermo Scientific™

Pierce™ Streptavidin Agarose Columns, 1 mL

Thermo Scientific Pierce Streptavidin Agarose Columns are 5-mL capacity columns containing 1 mL streptavidin agarose, for use in a varietyRead more
Have Questions?
Catalog NumberQuantity
203515 Columns
Catalog number 20351
Price (MXN)
-
Quantity:
5 Columns
Thermo Scientific Pierce Streptavidin Agarose Columns are 5-mL capacity columns containing 1 mL streptavidin agarose, for use in a variety of biotin-binding affinity purification methods

Features of Streptavidin Agarose:

• Streptavidin—purified recombinant streptavidin protein (53kDa, near-neutral pI); tetrameric with four biotin-binding site per molecule
• Agarose resin—support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
• Inert and stable—streptavidin is immobilized by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
• Standard capacity—this variety of beads has a normal load of immobilized streptavidin, providing a binding capacity of 1 to 3 mg of biotinylated BSA per mL of resin

Properties of crosslinked 6% beaded agarose (CL-6B):
• Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
• Average Particle Size: 45 to 165 microns
• Exclusion Limit: 10,000 to 4,000,000 daltons
• Maximum Volumetric Flow Rate: approx. 1 mL/minute (for 1 cm diameter column)
• Maximum Linear Velocity: 30 cm per hour
• Maximum Pressure: less than 25psi (1.5 bar)

Applications:

• Purification of membrane antigens in conjunction with biotinylated monoclonal antibodies
• Isolation of cell-surface proteins that have been labeled with a cleavable biotinylation reagent
• Isolation of cell-surface glycoproteins using biotinylated Concanavalin A
• Recovery of single-stranded biotinylated DNA for dideoxy sequencing
• Streptavidin-coated plates used for direct solid-phase sequencing
• In vitro mutagenesis with streptavidin-coated plates
• DNA fingerprinting with streptavidin-coated plates

Pierce Streptavidin Agarose consists of purified recombinant streptavidin that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose. Streptavidin is a tetrameric biotin-binding protein. The agarose beads have physical and chemical properties that enable them to be used in a variety of batch- or column-type affinity procedures. Biotinylated antibodies, proteins, peptides, nucleic acids and other molecules or interaction complexes can be captured, immunoprecipitated, removed or purified from samples using this streptavidin resin.

Streptavidin is a tetrameric protein containing four biotin-binding sites similar to avidin. The native protein from Streptomyces avidinii is carbohydrate-free, has an acidic pI of 5.5 and a mass of 75 kDa. The recombinant protein used to manufacture Thermo Scientific Pierce Streptavidin Resins has a near-neutral isoelectric point and a mass of 53kDa. Streptavidin generally has less nonspecific binding than avidin (from chicken egg white) because of the absence of carbohydrates.

Related Products
Pierce™ Streptavidin Agarose
Pierce™ High Capacity Streptavidin Agarose
Pierce™ Streptavidin Plus UltraLink™ Resin
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)5 mL
Column TypeAffinity, Agarose Resin
FormatSpin Column
Protein FormRecombinant
Quantity5 Columns
Stationary PhaseStreptavidin
Sufficient ForBinding 15 to 28 μg Biotin Per Column (1 to 3 mg Biotinylated BSA Per Column)
TargetBiotin-Tagged Molecule
Particle Size45 to 165 μm
Product LinePierce
TypeAgarose
Unit SizeEach
Contents & Storage
Contents: 5 × 1 mL prepacked columns with twist-off bottom tab and accessory pack containing five white tips.
Storage: Upon receipt store at 4°C. Do not freeze the resin.
Product is shipped at ambient temperature.

Frequently asked questions (FAQs)

How many biotin binding-sites are there per streptavidin molecule?

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.