Nalgene™ Narrow-Mouth HDPE Economy Bottles
Nalgene™ Narrow-Mouth HDPE Economy Bottles
Nalgene™ Narrow-Mouth HDPE Economy Bottles
Nalgene™ Narrow-Mouth HDPE Economy Bottles
Nalgene™ Narrow-Mouth HDPE Economy Bottles
Nalgene™ Narrow-Mouth HDPE Economy Bottles
Nalgene™ Narrow-Mouth HDPE Economy Bottles
Thermo Scientific™

Nalgene™ Narrow-Mouth HDPE Economy Bottles

Contain samples and daily working solutions like buffers and bench reagents with these HDPE economy bottles, suitable for lighter-duty, general purpose applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2089-0016500 mL28 mm
2089-000260 mL20 mm
2089-0004125 mL24 mm
2089-0008250 mL24 mm
2089-00321 L38-430 mm
Catalog number 2089-0016
Price (USD)
129.67
Online exclusive
148.19
Save 18.52
Case of 48
Capacity (Metric):
500 mL
Closure Size:
28 mm
Price (USD)
129.67
Online exclusive
148.19
Save 18.52
Case of 48

Contain samples and daily working solutions like buffers and bench reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Economy Bottles, lightweight versions of the original Nalgene Lab Quality Bottle. Suitable for lighter-duty, general purpose applications, these high-density polyethylene bottles are leakproof† when used with Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

  • Linerless polypropylene caps ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions
  • Recommended for sample collection and storage of daily working solutions
  • Bottles participate in My Green Labs' ACT Label program for Accountability, Consistency and Transparency around product sustainability
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Recycling Information
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
ACT LabelYes
AutoclavableNot Autoclavable
Capacity16 oz., 500 mL
Capacity (English)16 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorNatural
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
DescriptionNarrow-Mouth Economy Bottle
Diameter (Metric)73 mm
Diameter (Metric) Inner Neck21 mm
DisposableReusable
For Use With (Application)Lighter-Duty, General Purpose
Height (English)2.4 in.
Height (Metric)170 mm
Neck Finish28 mm
No. per Case48
No. per Pack48
Product TypeNarrow Mouth Economy Bottle
ShapeRound
Through the Closure DispensingNo
Capacity (Metric)500 mL
Closure IncludedYes
ColorTranslucent
MaterialHDPE
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 48

Frequently asked questions (FAQs)

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.