Pierce™ Gentle Ag/Ab Binding Buffer, pH 8.0
Thermo Scientific™

Pierce™ Gentle Ag/Ab Binding Buffer, pH 8.0

Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
210201 L
210123.75 L
Catalog number 21020
Price (USD)
191.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
191.00
Each
Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads. The Gentle Ag/Ab Binding and Elution Buffer system uses near-neutral, high-salt conditions for elution, enabling even sensitive and labile antibodies (or other proteins) to be eluted from an affinity system without denaturation and inactivation. The phosphate-free binding buffer in this system is a necessary substitute for the Protein A binding buffer (pH 8), which precipitates with the Gentle Ag/Ab Elution Buffer.

Shared features of all Thermo Scientific Pierce IgG Binding and Elution Buffers:

• Ready to use—carefully formulated and prefiltered 1X solutions
• Three binding buffers—optimized for Protein A, Protein G or Protein A/G
• Two elution buffer options—acidic (pH 2.8) or high salt (near-neutral)
• Versatile—elution buffers are appropriate for antibody-antigen affinity purification

Products available for each antibody purification method:
• Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
• Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
• Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
• IgG Elution Buffer: pH 2.8; amine-based
• Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
• Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

Related Products
Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialBinding Buffer
Recommended StorageUpon receipt store at 4°C
Physical FormLiquid
Product LinePierce, Pierce, Pierce, Pierce
Quantity1 L
pH8
Unit SizeEach