Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6
Thermo Scientific™

Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6

Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21027500 mL
210133.75 L
Catalog number 21027
Price (USD)
160.65
Online exclusive
181.00
Save 20.35
Each
Quantity:
500 mL
Price (USD)
160.65
Online exclusive
181.00
Save 20.35
Each
Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads. The Gentle Ag/Ab Binding and Elution Buffer system uses near-neutral, high-salt conditions for elution, enabling even sensitive and labile antibodies (or other proteins) to be eluted from an affinity system without denaturation and inactivation. The phosphate-free binding buffer in this system is a necessary substitute for the Protein A binding buffer (pH 8), which precipitates with the Gentle Ag/Ab Elution Buffer.

Shared features of all Thermo Scientific Pierce IgG Binding and Elution Buffers:

• Ready to use—carefully formulated and prefiltered 1X solutions
• Three binding buffers—optimized for Protein A, Protein G or Protein A/G
• Two elution buffer options—acidic (pH 2.8) or high salt (near-neutral)
• Versatile—elution buffers are appropriate for antibody-antigen affinity purification

Products available for each antibody purification method:
• Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
• Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
• Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
• IgG Elution Buffer: pH 2.8; amine-based
• Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
• Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

Related Products
Pierce™ IgG Elution Buffer
Pierce™ Gentle Ag/Ab Binding Buffer, pH 8.0
Pierce™ Gentle Ag/Ab Binding and Elution Buffer Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialElution Buffer
Recommended Storage4°C
Physical FormLiquid
Product LinePierce
Quantity500 mL
pH6.6
Unit SizeEach

Frequently asked questions (FAQs)

After immobilizing my antibody, the first affinity purification of my protein worked well, but the protein failed to bind to the antibody during the second round of purification. What did I do wrong?

Low pH elution is the most commonly used method of elution for affinity purification, however, it could lead to denaturation of some antibodies which will negatively affect subsequent antigen binding. To prevent this from happening, after protein purification and column regeneration, immediately wash the column with neutralization or binding buffer and store at 4 degrees C in buffer containing an antimicrobial agent such as sodium azide.

Alternatively, use a near-neutral, high salt elution buffer such as Pierce Gentle Ag/Ab Elution Buffer (Cat. No. 21013, 21027, or 21030). For alternative elution buffers, please refer to this Tech Tip (https://tools.thermofisher.com/content/sfs/brochures/TR0027-Elution-conditions.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How many purifications can be performed using the same affinity column?

This varies with the stability of the immobilized protein and the type of elution buffer used. Columns can typically be reused at least 10 times without significant loss of activity. If the immobilized molecule is sensitive to low pH, consider using our Pierce Gentle Ag/Ab Elution Buffer (Cat. No. 21013, 21027 or 21030). For alternative elution buffers, please refer to this Tech Tip(http://tools.thermofisher.com/content/sfs/brochures/TR0027-Elution-conditions.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.