Pierce™ IgG Elution Buffer
Thermo Scientific™

Pierce™ IgG Elution Buffer

Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
210041 L
210093.75 L
Catalog number 21004
Price (USD)
116.65
Online exclusive
134.00
Save 17.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
116.65
Online exclusive
134.00
Save 17.35
Each
Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads. The Pierce IgG Elution Buffer provides efficient dissociation and elution of antibodies from all three IgG-binding proteins, including Protein L. This low-pH elution buffer is also effective for most antibody-antigen or protein-protein affinity purification systems.

Shared features of all Thermo Scientific Pierce IgG Binding and Elution Buffers:

• Ready to use—carefully formulated and prefiltered 1X solutions
• Three binding buffers—optimized for Protein A, Protein G or Protein A/G
• Two elution buffer options—acidic (pH 2.8) or high salt (near-neutral)
• Versatile—elution buffers are appropriate for antibody-antigen affinity purification

Products available for each antibody purification method:
• Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
• Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
• Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
• IgG Elution Buffer: pH 2.8; amine-based
• Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
• Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

Related Products
Pierce™ IgG Elution Buffer
Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialElution Buffer
Recommended Storage4°C
Physical FormLiquid
Product LinePierce
Quantity1 L
pH2.8
Unit SizeEach