EZ-Link™ Pentylamine-Biotin
Thermo Scientific™

EZ-Link™ Pentylamine-Biotin

Thermo Scientific EZ-Link Pentylamine-Biotin is biotin that has been conjugated to pentylenediamine to add a short, amine-terminated spacer to itsRead more
Have Questions?
Catalog NumberQuantity
2134550 mg
Catalog number 21345
Price (MXN)
-
Quantity:
50 mg
Request bulk or custom format
Thermo Scientific EZ-Link Pentylamine-Biotin is biotin that has been conjugated to pentylenediamine to add a short, amine-terminated spacer to its valeric acid group, providing a simple amine-modified compound for different applications.

Features of EZ-Link Pentylamine-Biotin:

• Biotinylation—label molecules and surfaces for assay or affinity purification methods involving avidin or streptavidin probes and resins
• Amine-activated—primary amine can be crosslinked to proteins and material surfaces using EDC and other crosslinkers
• Medium length—spacer arm (total length added to target) is 18.9 angstroms, representing a 6-atom extension of the native biotin valeric acid (and its conversion to a primary amine)

Pentylamine Biotin is a compound formed by modification of the valeric acid side chain of biotin with pentane-1,5-diamine. The compound contains terminal primary amine (-NH2), which provides a functional handle for derivatization or conjugation to proteins, surfaces and other molecules. Carbodiimide (EDC) and NHS-ester crosslinker chemistries are most often utilized for covalent modifications involving amino-biotin molecules. The compound is also useful as an amine-alternative to native (carboxylate) biotin for assays and methods that require a biotin standard or blocking step.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

Amino-biotin compounds can be conjugated to functional groups of proteins and other molecules in a variety of ways. The most common method is to crosslink the terminal primary amine to carboxyl groups using . Carboxyl groups (-COOH) occur in aspartate or glutamate residues and the carboxy-terminus of polypeptides. When activated with EDC (Part No. 22980), carboxylates react with amino (—NH2) groups to form amide bonds. Carboxylate molecules and surface materials can be pre-activated using EDC with Sulfo-NHS (Part No. 24510) for subsequent reaction to primary amines (see NHS-ester Chemistry).

Applications:
• Used for colorimetric assays for Factor XIII and cellular transglutaminase
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Impermeant
Label TypeBiotin & Analogs
Product LineEZ-Link
Product TypePentylamine-Biotin
Quantity50 mg
Reactive MoietyAmine
Chemical ReactivityCarboxylic Acid, Ketone, Aldehyde
Label or DyeBiotin
SolubilityWater
SpacerShort
Unit SizeEach
Contents & Storage
Store at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for EZ-Link Pentylamine-Biotin?

EZ-Link Pentylamine-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (1)

Citations & References
Abstract
Isolation and characterization of antibody fragments selective for toxic oligomeric tau.
Authors:Tian H, Davidowitz E, Lopez P, He P, Schulz P, Moe J, Sierks MR
Journal:
PubMed ID:25616912
Oligomeric tau species are important in the onset and progression of Alzheimer's disease (AD), as they are neurotoxic and can propagate tau-tangle pathology. Therefore, reagents that selectively recognize different key morphologies of tau are needed to help define the role of tau in AD and related diseases. We utilized a ... More