EGS (ethylene glycol bis(succinimidyl succinate)) - FAQs
EGS (ethylene glycol bis(succinimidyl succinate)) - FAQs
View additional product information for EGS (ethylene glycol bis(succinimidyl succinate)) - FAQs (21565)
2 product FAQs found
Can you provide the shelf-life for EGS (ethylene glycol bis(succinimidyl succinate))?
EGS (ethylene glycol bis(succinimidyl succinate)) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can NHS-diazirine (Cat. No. 26167) be used for CHIP applications?
NHS-diazirine has not been tested for cross-linking in a ChIP application. EGS (Cat. No. 21565) and DSG (Cat. No. 20593) are not used by themselves in ChIP, but rather in combination with formaldehyde. While formaldehyde is good for crosslinking proteins that are in direct contact with DNA, it cannot trap proteins that may be bound in a complex with other proteins but are not in direct contact. Thus EGS or DSG can cross-link proteins to proteins that are in direct contact with DNA and are crosslinked to it by formaldehyde. See this reference for more information
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.