Search
View additional product information for BS3 (bis(sulfosuccinimidyl)suberate) - FAQs (21586, 21580, A39266)
3 product FAQs found
BS3 is the water-soluble analogue of DSS (i.e., DSS is the membrane-permeable analog of BS3). DSS is water-insoluble and membrane-permeable, so it can be used for both intracellular and cell surface crosslinking. BS3 is water-soluble and membrane-impermeable, so it can be used for cell-surface crosslinking.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
For crosslinking the antibody to Protein G Dynabeads, please refer to the crosslinking protocol with BS3 (bis(sulfosuccinimidyl)suberate) or Sulpha-DSS, which is an analogue of DSS. The only difference between these two molecules is that DSS is water insoluble and needs to be solubilized in DMSO before use. You can find the protocol at the following link.
Find additional tips, troubleshooting help, and resources within our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center
BS3 (bis(sulfosuccinimidyl)suberate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.