BS3 (bis(sulfosuccinimidyl)suberate)
BS3 (bis(sulfosuccinimidyl)suberate)
BS3 (bis(sulfosuccinimidyl)suberate)
BS3 (bis(sulfosuccinimidyl)suberate)
Thermo Scientific™

BS3 (bis(sulfosuccinimidyl)suberate)

Thermo Scientific Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.BS3 contains anRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2158050 mg
A3926610 x 2 mg
215861 g
Catalog number 21580
Price (USD)
179.00
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
179.00
Each
Thermo Scientific Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.

BS3 contains an amine-reactive N-hydroxysulfosuccinimide (NHS) ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxysulfosuccinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS-ester crosslinking reagents.

No-Weigh microtube handling:
Each of the eight microtubes contains 2 mg of crosslinker. Puncture foil with a pipette tip and add DMSO or DMF to the DSS or aqueous buffer to the BS3. Store the microtube strip in the foil pouch provided. Used microtubes may be cut from the unused microtubes and discarded.

Features of BS3:

• Reactive groups: sulfo-NHS ester (both ends)
• Reactive towards: amino groups (primary amines)
• Amine-reactive Sulfo-NHS ester reacts rapidly with any primary amine-containing molecule
• Water-soluble; compare with DSS
• Membrane-impermeable, allowing for cell surface labeling
• High-purity, crystalline reagent can be used to create high-purity crosslinked conjugates

Because it contains the hydrophilic sulfonyl moiety, BS3 crosslinker is soluble up to ∼100 mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure. DSS, the non-water soluble analog of BS3 is also available for applications that require a less hydrophilic crosslinker (e.g., to effect intracellular crosslinking). DSS and BS3 have essentially identical crosslinking activity toward primary amines.

Properties of BS3:
• Alternative names: Sulfo-DSS
• Molecular formula: C16H18N2O14S2Na2
• Molecular weight: 572.43
• Spacer arm length: 11.4 Å (8 atoms)
• CAS number: 82436-77-9
• Reactive groups: NHS esters, react with primary amines at pH 7.0-9.0

Applications:
• Crosslink cell-surface proteins prior to cell lysis and immunoprecipitation
• Identify receptor-ligand interactions by chemical crosslinking
• 'Fix' protein interactions to allow identification of weak or transient protein interactions
• Identify near-neighbor protein interactions
• Protein crosslinking to create bioconjugates via single-step reactions
• Immobilize proteins onto amine-coated surfaces

BS3 Specifications:
We manufacture BS3 to the highest specifications to produce the most specific bioconjugates, ensure the integrity of your data and to provide you with the highest degree of consistency. Each lot of BS3 is tested to meet the following minimum specifications:
• Purity: > 93% by quantitative NMR (the highest standard for crosslinker purity)
• Solubility: > 5.8 mg/mL in DI water, clear solution with no insoluble material

Product References
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Pierce™ Premium Grade BS3
DSS (disuccinimidyl suberate)
BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate)
BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)572.43
PEGylatedNo
Product LinePierce
Quantity50 mg
Reactive MoietySulfo-NHS Ester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length11.4 Å
Water SolubleYes
Chemical ReactivityAmine-Amine
CleavableNo
Crosslinker TypeHomobifunctional
FormatStandard, Single-use, Premium-grade
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at 4°C.

Frequently asked questions (FAQs)

What is the difference between DSS and BS3 crosslinkers?

BS3 is the water-soluble analogue of DSS (i.e., DSS is the membrane-permeable analog of BS3). DSS is water-insoluble and membrane-permeable, so it can be used for both intracellular and cell surface crosslinking. BS3 is water-soluble and membrane-impermeable, so it can be used for cell-surface crosslinking.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the protocol for crosslinking IgG to Dynabeads from the Dynabeads Protein G Immunoprecipitation Kit (Cat. No. 10007D) using DSS (disuccinimidyl suberate)?

For crosslinking the antibody to Protein G Dynabeads, please refer to the crosslinking protocol with BS3 (bis(sulfosuccinimidyl)suberate) or Sulpha-DSS, which is an analogue of DSS. The only difference between these two molecules is that DSS is water insoluble and needs to be solubilized in DMSO before use. You can find the protocol at the following link.

Find additional tips, troubleshooting help, and resources within our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center

Can you provide the shelf-life for BS3 (bis(sulfosuccinimidyl)suberate)?

BS3 (bis(sulfosuccinimidyl)suberate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (8)

Citations & References
Abstract
Identification of mannose receptor as receptor for hepatocyte growth factor ß-chain: novel ligand-receptor pathway for enhancing macrophage phagocytosis.
Authors:Ohnishi H, Oka K, Mizuno S, Nakamura T
Journal:J Biol Chem
PubMed ID:22354962
'Hepatocyte growth factor (HGF), a heterodimer composed of the a-chain and ß-chain, exerts multifunctional actions for tissue repair and homeostasis via its receptor, MET. HGF is cleaved by proteases secreted from inflammatory cells, and NK4 and ß-chain remnant (HGF-ß) are generated. Here, we provide evidence that HGF-ß binds to a ... More
Anti-human activin receptor-like kinase 1 (ALK1) antibody attenuates bone morphogenetic protein 9 (BMP9)-induced ALK1 signaling and interferes with endothelial cell sprouting.
Authors:van Meeteren LA, Thorikay M, Bergqvist S, Pardali E, Stampino CG, Hu-Lowe D, Goumans MJ, ten Dijke P
Journal:J Biol Chem
PubMed ID:22493445
'Genetic and molecular studies suggest that activin receptor-like kinase 1 (ALK1), a transforming growth factor ß (TGF-ß) type I receptor, and endoglin, a TGF-ß co-receptor, play an essential role in vascular development and pathological angiogenesis. Several agents that interfere with ALK1 and endoglin function are currently in clinical trials for ... More
Tight complex formation between Cosmc chaperone and its specific client non-native T-synthase leads to enzyme activity and client-driven dissociation.
Authors:Aryal RP, Ju T, Cummings RD
Journal:J Biol Chem
PubMed ID:22416136
'The interaction of the endoplasmic reticulum molecular chaperone Cosmc with its specific client T-synthase (Core 1 ß1-3-galactosyltransferase) is required for folding of the enzyme and eventual movement of the T-synthase to the Golgi, but the mechanism of interaction is unclear. Here we show that the lumenal domain of recombinant Cosmc ... More
Resolving nitrogen-15 and proton chemical shifts for mobile segments of elastin with two-dimensional NMR spectroscopy.
Authors:Ohgo K, Niemczura WP, Seacat BC, Wise SG, Weiss AS, Kumashiro KK
Journal:J Biol Chem
PubMed ID:22474297
In this study, one- and two-dimensional NMR experiments are applied to uniformly (15)N-enriched synthetic elastin, a recombinant human tropoelastin that has been cross-linked to form an elastic hydrogel. Hydrated elastin is characterized by large segments that undergo "liquid-like" motions that limit the efficiency of cross-polarization. The refocused insensitive nuclei enhanced ... More
The tumor necrosis factor receptor stalk regions define responsiveness to soluble versus membrane-bound ligand.
Authors:Richter C, Messerschmidt S, Holeiter G, Tepperink J, Osswald S, Zappe A, Branschädel M, Boschert V, Mann DA, Scheurich P, Krippner-Heidenreich A,
Journal:Mol Cell Biol
PubMed ID:22547679
The family of tumor necrosis factor receptors (TNFRs) and their ligands form a regulatory signaling network that controls immune responses. Various members of this receptor family respond differently to the soluble and membrane-bound forms of their respective ligands. However, the determining factors and underlying molecular mechanisms of this diversity are ... More