Nalgene™ Wide-Mouth PPCO Economy Bottles with Closure, 30mL - FAQs
View additional product information for Nalgene™ Wide-Mouth PPCO Economy Bottles with Closure - FAQs (2187-0032, 2187-0004, 2187-0001)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.