DMEM, low glucose, pyruvate, HEPES, 10 x 500 mL - FAQs
View additional product information for DMEM, low glucose, pyruvate, HEPES - FAQs (22320030)
1 product FAQs found
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.