How long is Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) (Cat. No. 22322) functional group (maleimide) stable for after the amine targeted end is occupied?
The Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) (Cat. No. 22322) maleimide group is stable for 64 hours in 0.1 M sodium buffer, pH 7 at 4 degrees C.