Pierce™ 660nm Protein Assay Kit
Thermo Scientific™

Pierce™ 660nm Protein Assay Kit

The Pierce 660-nm Protein Assay is a ready-to-use, detergent- and reducing agent-compatible assay reagent to quickly measure total protein concentrationRead more
Have Questions?
Catalog NumberQuantity
22662450 mL
Catalog number 22662
Price (USD)
321.00
Each
Quantity:
450 mL
Request bulk or custom format
Price (USD)
321.00
Each
The Pierce 660-nm Protein Assay is a ready-to-use, detergent- and reducing agent-compatible assay reagent to quickly measure total protein concentration compared to a protein standard. The assay is more linear than Coomassie-based Bradford assays and compatible with higher concentrations of most detergents, reducing agents, and other commonly used reagents.

See all available protein assays ›

Features of the 660-nm Protein Assay Kit:
• Compatibility—works with a greater range of detergents and reducing agents than other dye-based assays
• Ready-to-use—single reagent with a simple mix-and-read protocol; no working reagent to prepare
• Linearity—produces standard curves that are more linear than with the Bradford assay method
• Assay format—assay may be performed in test tubes or microplates
• Sample volume—requires only 10 μL for microplate or 100 μL for the test tube procedures
• Storage—room temperature storage

The accessory Ionic Detergent Compatibility Reagent (IDCR) provides for even broader detergent compatibility, making this one of the only protein assays that is suitable for samples containing Laemmli SDS sample buffer with bromophenol blue. Although the Pierce 660-nm Protein Assay produces a higher level of protein-to-protein variation (37%) than other assays, such as the BCA Protein Assay, the simpler single-reagent format and broader substance compatibility make the Pierce 660-nm assay more convenient for many routine applications. The Pierce 660-nm Protein Assay can be performed in either a test tube or microplate format.

How the Pierce 660-nm Assay detects protein
The Pierce 660-nm Protein Assay is based on the binding of a proprietary dye-metal complex to protein in acidic conditions that causes a shift in the dye's absorption maximum, which is measured at 660 nm. The dye-metal complex is reddish-brown and changes to green upon protein binding. The color change is produced by deprotonation of the dye at low pH facilitated by interactions with positively charged amino acid groups in proteins. Therefore, the dye interacts mainly with basic residues in proteins, such as histidine, arginine, and lysine and to a lesser extent tyrosine, tryptophan, and phenylalanine.

The color produced in the assay is stable and increases in proportion to a broad range of increasing protein concentrations, even in the presence of detergents and reducing agents that would be incompatible with Bradford and BCA protein assays.

Related products
Pierce 660-nm Protein Assay Reagent
Ionic Detergent Compatibility Reagent for Pierce 660-nm Protein Assay Reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayProtein Assay Kit
DescriptionPierce 660nm Protein Assay Kit
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce
Product TypeProtein Quantitation Assay
Quantity450 mL
SpecificityNot Target-Specific
Sufficient For300 Tube Assays or 3000 Microplate Assays
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Sufficient For: 300 tube assays or 3000 microplate assays
• Pierce 660 nm Protein Assay Reagent, 450 mL
• Albumin Standard Pre-Diluted Set, 7 x 3.5 mL

Store the assay reagent at room temperature and the pre-diluted BSA standards at 4°C.

Frequently asked questions (FAQs)

Which Thermo Scientific protein assay is the best or the most reliable?

The choice of protein assay is dependent on preferences related to assay speed, accuracy and sensitivity, as well as interfering substances in the sample to be assayed. BCA has less protein-to-protein variation, is compatible with most detergents, and has larger working range. Pierce Bradford Plus Protein Assay Kit (Cat. No. 23236) is compatible with reducing sugars, is more sensitive and is faster and easier to use. For a comparison of different protein assays and compatible reagents, see our Tech Tip: Protein Quantitation Assay Compatibility Table (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

Can I use the Thermo Scientific 660 nm Protein Assay to quantify peptides?

The Thermo Scientific 660 nm Protein Assay can quantify peptides that are at least 2,500 Da if their compositions include amino acid residues that react with the dye-metal reagent (i.e., histidine, arginine, tyrosine, tryptophan and phenylalanine). For peptides smaller than 2,500 Da, use the Fluoraldehyde Reagent Solution (Cat. No. 26025), which detects amino groups.

Other peptide quantitation assays we offer are Thermo Scientific Pierce Quantitative Colorimetric Peptide Assay (Cat. No. 23275) and Thermo Scientific Pierce Quantitative Fluorometric Peptide Assay (Cat. No. 23290).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the linear range of the Thermo Scientific 660 nm Protein Assay?

The assay's linear range is 25-2,000 µg/mL using the test tube procedure and 50-2,000 µg/mL using the microplate procedure.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I measure the absorbance at a wavelength other than 660 nm when using the Thermo Scientific 660nm Protein Assay Kit?

If a 660nm filter is not available, the assay can be measured at wavelengths from 645 to 670nm; however, the assay linear range is 25-2,000 µg/mL and occurs only when the absorbance of the dye-protein complex is measured at 660 nm. Measuring the absorbance at wavelengths other than 660nm will result in a decrease of the assay's linear range and might increase the minimum detection level (i.e., decrease sensitivity).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What advantages does the Thermo Scientific 660nm Protein Assay have over other protein assays?

The Thermo Scientific 660 nm Protein Assay has the following advantages: • Room temperature stability of the assay reagent • A greater linear range than the coomassie-based Bradford assays • Compatibility with commonly used detergents and reducing agents • Compatibility with samples lysed in Laemmli sample buffer • Rapid mix-and-read protocol

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (13)

Citations & References
Abstract
A rapid and versatile method for the isolation, purification and cryogenic storage of Schwann cells from adult rodent nerves.
Authors:Andersen ND, Srinivas S, Piñero G, Monje PV
Journal:Sci Rep
PubMed ID:27549422
'We herein developed a protocol for the rapid procurement of adult nerve-derived Schwann cells (SCs) that was optimized to implement an immediate enzymatic dissociation of fresh nerve tissue while maintaining high cell viability, improving yields and minimizing fibroblast and myelin contamination. This protocol introduces: (1) an efficient method for enzymatic ... More
Overexpression of the Sorghum bicolor SbCCoAOMT alters cell wall associated hydroxycinnamoyl groups.
Authors:Tetreault HM, Scully ED, Gries T, Palmer NA, Funnell-Harris DL, Baird L, Seravalli J, Dien BS, Sarath G, Clemente TE, Sattler SE
Journal:PLoS One
PubMed ID:30289910
'Sorghum (Sorghum bicolor) is a drought tolerant crop, which is being developed as a bioenergy feedstock. The monolignol biosynthesis pathway is a major focus for altering the abundance and composition of lignin. Caffeoyl coenzyme-A O-methyltransferase (CCoAOMT) is an S-adenosyl methionine (SAM)-dependent O-methyltransferase that methylates caffeoyl-CoA to generate feruloyl-CoA, an intermediate ... More
Exosomes facilitate therapeutic targeting of oncogenic KRAS in pancreatic cancer.
Authors:Kamerkar S, LeBleu VS, Sugimoto H, Yang S, Ruivo CF, Melo SA, Lee JJ, Kalluri R
Journal:Nature
PubMed ID:28607485
'The mutant form of the GTPase KRAS is a key driver of pancreatic cancer but remains a challenging therapeutic target. Exosomes are extracellular vesicles generated by all cells, and are naturally present in the blood. Here we show that enhanced retention of exosomes, compared to liposomes, in the circulation of ... More
Mechanisms of PKC-Mediated Enhancement of HIF-1a Activity and its Inhibition by Vitamin K2 in Hepatocellular Carcinoma Cells.
Authors:Xia J, Ozaki I, Matsuhashi S, Kuwashiro T, Takahashi H, Anzai K, Mizuta T
Journal:Int J Mol Sci
PubMed ID:30813635
'Hypoxia-inducible factor 1 (HIF-1) plays important roles in cancer cell biology. HIF-1a is reportedly activated by several factors, including protein kinase C (PKC), in addition to hypoxia. We investigated the role of PKC isoforms and the effects of vitamin K2 (VK2) in the activation process of HIF-1a. Human hepatocellular carcinoma ... More
Genetic resistance to DEHP-induced transgenerational endocrine disruption.
Authors:Stenz L, Rahban R, Prados J, Nef S, Paoloni-Giacobino A
Journal:PLoS One
PubMed ID:31181066
'Di(2-ethylhexyl)phthalate (DEHP) interferes with sex hormones signaling pathways (SHP). C57BL/6J mice prenatally exposed to 300 mg/kg/day DEHP develop a testicular dysgenesis syndrome (TDS) at adulthood, but similarly-exposed FVB/N mice are not affected. Here we aim to understand the reasons behind this drastic difference that should depend on the genome of ... More