Thermo Scientific Pierce MMTS (methyl methanethiosulfonate) is a small compound that reversibly blocks cysteines and other sulfhydryl groups, enabling theRead more
Have Questions?
Catalog Number
Quantity
23011
200 mg
Catalog number 23011
Price (USD)
157.00
Each
Quantity:
200 mg
Request bulk or custom format
Price (USD)
157.00
Each
Thermo Scientific Pierce MMTS (methyl methanethiosulfonate) is a small compound that reversibly blocks cysteines and other sulfhydryl groups, enabling the study of enzyme activation and other protein functions.
Features of MMTS (methyl methanethiosulfonate):
• Converts sulfhydryl groups on cysteine side chains into -S-S-CH3 • Reaction is reversible with DTT or TCEP, restoring the free sulfhydryl • Used to modify thiol groups in creatine kinase
MMTS (methyl methanethiosulfonate) is a sulfhydryl-reactive compound that can reversibly sulfenylate thiol-containing molecules. Reacting MMTS with reduced sulfhydryls (-SH) results in their modification to dithiomethane (-S-S-CH3). Treatment with reducing agents, such as dithiothreitol (DTT), 2-mercaptoethanol (2-ME) or Tris(2-carboxyethyl) phosphine hydrochloride (TCEP), will cleave the disulfide groups to restore the original sulfhydryl. MMTS is commonly used to study biochemical pathways involving thiol-dependent enzymes.