Search
View additional product information for StemPro™ EZPassage™ Disposable Stem Cell Passaging Tool - FAQs (23181010)
1 product FAQs found
The tool has 230 micrometers between each blade. We don't recommend using the tool on anything smaller than a 6-well plate. As far as the thickness at the base of each blade relative to the distance between the blade's edges, we don't have data on that.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.