Pierce™ Bovine Serum Albumin Standard Ampules, 2 mg/mL
Thermo Scientific™

Pierce™ Bovine Serum Albumin Standard Ampules, 2 mg/mL

Thermo Scientific Pierce BSA Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in자세히 알아보기
Have Questions?
카탈로그 번호수량
2320910 x 1 mL
카탈로그 번호 23209
제품 가격(KRW)
117,000
온라인 행사
Ends: 31-Dec-2026
129,000
할인액 12,000
Each
수량:
10 x 1 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
117,000
온라인 행사
Ends: 31-Dec-2026
129,000
할인액 12,000
Each
Thermo Scientific Pierce BSA Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in total protein assays.

Features of BSA Protein Assay Standards:

• Convenient—1 mL ampules
• Universal—recognized as the industry standard for protein quantitation in colorimetric protein assays
• Pure and stable—supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable
• Accurate and consistent—precisely formulated at 2.00 +/-0.03 mg/mL compared to an NIST reference

These bovine serum albumin (BSA) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BSA is the universally accepted reference protein for total protein quantitation. The albumin standard is precisely formulated at 2 mg/mL in an ultrapure 0.9% sodium chloride (saline) solution. The concentration of the stock solution is calibrated by direct comparison to purified BSA (Fraction V) from the National Institute of Standards and Technology (NIST).

Applications:
• Protein assay quantitation standard (BCA Protein Assay, Coomassie-Bradford Assay, etc.)
• Protein recovery control for desalting and other column procedures
• General calibration of spectrophotometer UV-lamp (absorbance at 280nm)

Selection of a protein standard is potentially the greatest source of error in any protein assay. The best choice for a standard is a purified, known concentration of the most abundant protein contained in the samples being tested. Often, a highly purified, known concentration of the protein of interest is not available or it is too expensive to use as the standard, or the sample itself is a mixture of many proteins (e.g., cell lysate). In such cases, the best standard is one that will produce a normal (i.e., average) color response curve with the selected protein assay method and is readily available to any researcher. BSA is such a protein, and the Pierce Albumin Standards are the most convenient source of ready-to-use BSA standard.

For greatest accuracy in estimating total protein concentration in unknown samples, it is essential to include a standard curve each time the assay is performed. This is particularly true for the protein assay methods that produce non-linear standard curves. Deciding on the number of standards and replicates used to define the standard curve depends upon the degree of non-linearity in the standard curve and the degree of accuracy required. In general, fewer points are needed to construct a standard curve if the color response is linear. Typically, standard curves are constructed using at least two replicates for each point on the curve.

For added safety when opening glass ampules, consider using our Ampule Breakers, which are disposable safety devices that protect the fingers when breaking open a glass ampule.

Related Products
Pierce™ Bovine Serum Albumin Standard Pre-Diluted Set
For Research Use Only. Not for use in diagnostic procedures.
사양
어세이Protein Standards
설명Pierce Bovine Serum Albumin Standard Ampules, 2mg/mL
용도(애플리케이션)Internal Standard
제품라인Pierce
제품 유형Protein Quantitation Assay
수량10 x 1 mL
특이성Not Target-Specific
Unit SizeEach
구성 및 보관
Store at room temperature.

자주 묻는 질문(FAQ)

Can you provide the shelf-life for the Pierce Bovine Serum Albumin Standard, 2 mg/mL?

The Pierce Bovine Serum Albumin Standard, 2 mg/mL is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can you provide the shelf life for Pierce Bovine Serum Albumin Standard Ampules, 2 mg/mL?

Pierce Bovine Serum Albumin Standard Ampules, 2 mg/mL are covered under our general 1-year warranty and are guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (room temperature). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

When using a NanoDrop spectrophotometer, what is the difference between controls versus standards?

NanoDrop CF-1 Calibration Fluid is the only photometric standard available to assess the accuracy of the NanoDrop spectrophotometer pedestal. If the control sample shows unexpected results the user should confirm the calibration of the instrument with the CF-1 fluid.
Some control examples include:
- Nucleic Acids: Fermentas GeneRuler Express DNA Ladder 100-5000 bp (Cat. No. SM1551)
- Protein: Pierce Bovine Serum Albumin Standard Ampules, 2 mg/mL (Cat. No. 23209)

How should I open the glass BSA standard ampules?

There are several ampule-breaking devices sold through various suppliers such as Fisher Scientific (see https://www.fishersci.com/us/en/catalog/search/products?keyword=ampule+breaker&nav=) that can be used.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I re-use diluted BSA standards for subsequent protein assays?

The challenge of diluted BSA standards is that they need to be protected from microbial growth, evaporation and protein precipitation. These will result in artificially high or low protein concentration values. Including additional sodium azide with the diluted protein standards to maintain a final concentration of 0.05% will help prevent microbial growth. The shelf life of diluted standards can be extended if they are sealed and kept refrigerated.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.