Will the reducing agent in my sample interfere with the Glycoprotein Carbohydrate Estimation assay?
Yes. Reducing agents interfere with this assay. Please see the list of compatibility of other commonly used reagents below:
Compatible: EDTA, Urea (<1.0M), Guanidine HCl (<1.0M), DMSO (<10%), PBS
Not Compatible: Reducing Agents, Tris, Glycerol