Pierce™ Peroxide Assay Reagent C
Thermo Scientific™
Pierce™ Peroxide Assay Reagent C
TThermo Scientific Pierce Peroxide Assay Reagent C is a formulation consisting of 125μM xylenol orange in methanol with BHT. ThisRead more
Have Questions?
Catalog NumberQuantity
2328325 mL
Catalog number 23283
Price (USD)
174.00
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
174.00
Each
TThermo Scientific Pierce Peroxide Assay Reagent C is a formulation consisting of 125μM xylenol orange in methanol with BHT. This formulation is one of the component of Quantitative Peroxide Assay Kit used for detecting and measuring hydrogen peroxide levels based on the oxidation of ferrous to ferric ion in the presence of xylenol orange, present in the biological and other liquid samples.
Features of Peroxide Assay Reagent B:
  • Convenient – no proprietary reagents involved and remove the variability, difficulty and added expense of formulating the reagents yourself
  • Lipid-compatible formulations – the lipid-compatible formulation provides the best results with lipid-containing samples without first extracting the lipids;however, because this kit does not contain sorbitol, the assay is less sensitive.
Applications of Pierce Quantitative Peroxide Assay Kits:
  • General hydrogen peroxide (H2O2) measurement in protein samples
  • Quantitation of peroxides in detergents
  • Assessment of the effects of peroxides on cellular activity
  • Determination of protein glycation; Measurement of peroxide accumulation in lipids
In these assay kits, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With the aqueous-compatible formulation, peroxide first reacts with sorbitol, converting it to a peroxyl radical, which in turn initiates Fe2+ oxidation to Fe3+. In the lipid compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560nm. Peroxide levels in test samples can be determined by calculation from the know extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.Most proteins do not interfere with these assays, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.
Related Products
Pierce™ Quantitative Peroxide Assay Kit (Aqueous)
Pierce™ Quantitative Peroxide Assay Kit (Lipid)
Pierce™ Peroxide Assay Reagent A
Pierce™ Peroxide Assay Reagent B
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayPeroxide Assay Reagent C
DescriptionPierce Peroxide Assay Reagent C
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce
Product TypeQuantitative Peroxide Assay
Quantity25 mL
SpecificityNot Target-Specific
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.