Pierce™ Quantitative Peroxide Assay Kit (Aqueous
Thermo Scientific™

Pierce™ Quantitative Peroxide Assay Kit (Aqueous

Detect and measure hydrogen peroxide levels (H2O2) in biological samples using an iron (Fe) and xylenol orange (XO) reagent for microplates or tubes.

Have Questions?
Catalog NumberQuantity
23280100 mL
Catalog number 23280
Price (USD)
271.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
271.00
Each

The Thermo Scientific Pierce Quantitative Peroxide Assay Kit (aqueous-compatible formulation) detects and measures hydrogen peroxide levels in biological samples using an iron and xylenol orange (XO) reagent.

This aqueous-compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange. The kit's formulation includes sorbitol, which provides sensitivity enhancement. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects. This kit provides for maximum sensitivity with non-lipid samples.

Features of the Quantitative Peroxide Assay Kit:

  • Peroxides—detects and measures hydrogen peroxide (H2O2) levels in biological and other liquids samples
  • Convenient—no proprietary reagents involved but the ready-to-use kit removes the variability, difficulty and added expense of formulating the reagents yourself

Applications:

  • General hydrogen peroxide (H2O2) measurement in protein samples
  • Quantitation of peroxides in detergents
  • Assessment of the effects of peroxides on cellular activity
  • Determination of protein glycation; Measurement of peroxide accumulation in lipids

In this assay, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With this aqueous-compatible formulation, peroxide first reacts with sorbitol, converting it to a peroxyl radical, which in turn initiates Fe2+ oxidation to Fe3+. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560nm. Peroxide levels in test samples can be determined by calculation from the known extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.

Related Products

Pierce™ Quantitative Peroxide Assay Kit (Lipid) (Cat. No. 23285)

Pierce™ Peroxide Assay Reagent A (Cat. No. 23281)

Pierce™ Peroxide Assay Reagent B (Cat. No. 23282)

Pierce™ Peroxide Assay Reagent C (Cat. No. 23283)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayQuantitative Peroxide Assay
DescriptionPierce Quantitative Peroxide Assay Kit (Aqueous)
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce
Product TypeQuantitative Peroxide Assay
Quantity100 mL
SpecificityNot Target-Specific
Sufficient For100 Tube Assays or 500 Microplate Assays
Detection MethodAbsorbance
Unit SizeEach
Contents & Storage
Sufficient For: 100 tube assays or 500 microplate assays with non-lipid samples
• Reagent A, 1 mL
• Reagent B, 2 x 50 mL

Store at 4°C.

Frequently asked questions (FAQs)

Do you offer any protein assays that are protein specific?

We offer several “specialty” protein assays including: Protease assay kits (Cat. Nos. 23263 and 23266), Glycoprotein Carbohydrate Estimation Kit (Cat. No. 23260), Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270), Quantitative Peroxide assay kits (Cat. Nos. 23280 and 23285), and Easy-Titer Assay kits for IgG and IgM.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.