Easy-Titer™ Human IgG (H+L) Assay Kit - FAQs
Easy-Titer™ Human IgG (H+L) Assay Kit - FAQs
View additional product information for Easy-Titer™ Human IgG (H+L) Assay Kit - FAQs (23310)
6 product FAQs found
Will the Easy-Titer Antibody Assay kits cross-react with other species or immunoglobulin types?
No. The level of cross-reaction is neglible for common mixtures that we have tested.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Will fetal bovine serum (FBS) in a sample interfere with the Easy-Titer Antibody assay?
No, these assay use beads with target-specific antibodies, so off-target immunoglobulins have little or no effect on assay performance.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I am having problems with my standard curve in the Easy-Titer Antibody Assay. What do I need to do?
Make sure that you are mixing the samples vigorously. We set our shaker near the highest setting for this step. For best results, the beads must be fully dispersed.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
How do I pipette small volumes in the Easy-Titer Antibody Assay?
The assay method requires many-fold dilution of sample (so that the target immunoglobulin is at 5-500 ng/mL). To obtain accurate results, this dilution must be done carefully. Use properly matched pipette sizes (e.g., do not use a 200 µl pipette to measure 10 µl) and make the dilution in two or three steps (e.g., 10-fold times 10-fold)
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Do the Easy-Titer Antibody Assay kits come with a microplate?
No. Any standard, clear 96-well microplate can be used for this assay method.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can the Easy-Titer Antibody Assay kits be used with cell culture supernatant?
Yes. Samples are diluted at least 20-fold (usually much more) in the assay dilution buffer, rendering them fully compatible with the bead agglutination method.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.