Search
Buscar
View additional product information for 8M Guanidine-HCl Solution - FAQs (24115)
1 product FAQs found
There is no expiration date for the 8M Guanidine-HCl Solution (Cat. No. 24115). Stability is not affected by opening the bottle.