Nalgene™ Fluorinated HDPE Solvent Wash Bottle - FAQs
View additional product information for Nalgene™ Fluorinated HDPE Solvent Wash Bottle - FAQs (2421-0500PK)
1 product FAQs found
To what level are your FLPE fluorinated containers processed?
All of our fluorinated products are processed to Level 5.