I used the Thermo Scientific 6xHis Protein Tag Stain Reagent Set to detect my 6xHis-tagged protein but am also detecting non-tagged proteins. Why is this?
This is likely due to weak cross-reaction staining of proteins containing histidine clusters. Here are our recommendations:
- Wash gel for additional time in water (step 5.)
- Slightly decrease staining time (step 2.)
- Adjust exposure time and other settings to minimize weak, non-specific staining.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
- Wash gel for additional time in water (step 5.)
- Slightly decrease staining time (step 2.)
- Adjust exposure time and other settings to minimize weak, non-specific staining.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.