Pierce™ Reversible Protein Stain Kit for Nitrocellulose Membranes
Thermo Scientific™

Pierce™ Reversible Protein Stain Kit for Nitrocellulose Membranes

The Pierce Reversible Protein Stain Kit for Nitrocellulose Membranes is a rapid and sensitive alternative to Ponceau S stain forRead more
Have Questions?
Catalog NumberQuantity
245801.5 L
Catalog number 24580
Price (USD)
217.00
Each
Quantity:
1.5 L
Request bulk or custom format
Price (USD)
217.00
Each
The Pierce Reversible Protein Stain Kit for Nitrocellulose Membranes is a rapid and sensitive alternative to Ponceau S stain for protein detection on nitrocellulose membranes after transfer from polyacrylamide gels.

Features:
• Sensitive—high-avidity, total-protein stain; lower limit of detection equals 25 to 50 ng per band
• Specific—detects only protein; does not bind or interact with other electrophoresis or sample components
• Protocol—stain in less than five minutes; completely erase and reverse staining in less than 15 minutes
• Stability—components are stable at room temperature, saving refrigerator space and eliminating equilibration steps

Compare all membrane stains ›

This kit for membrane staining uses a nondestructive, reversible, reliable, and sensitive method to stain and detect proteins on nitrocellulose membranes. The lower limit of detection with this method is 25 to 50 ng per band (at least five times more sensitive than traditional Ponceau S staining). The stain has minimal nonspecific interactions with the membrane surface and protein-transfer reagents. The staining protocol is simple and quick and results in turquoise-blue bands that do not fade and are easily photographed for future reference. The stain can be easily reversed in less than 15 minutes. Subsequent western blot detection is unaffected because the stain does not alter the protein and is completely removed. In addition, the stain is compatible with N-terminal sequence analysis of proteins excised and eluted from membrane.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Reversible Protein Stain Kit for Nitrocellulose Membranes
Detection LocationIn-Blot Detection
Detection MethodColorimetric
Quantity1.5 L
Target MoleculeProtein
Label or DyeProprietary Mix
Product LinePierce
Product TypeReversible Protein Stain Kit
Unit SizeEach
Contents & Storage
Sufficient For: 10 nitrocellulose membranes (8 cm x 8 cm) from mini gels (SDS-PAGE)
• Reversible Stain, 250 mL
• Destain, 2 x 500 mL
• Stain Eraser, 250 mL

Store at room temperature

Frequently asked questions (FAQs)

How can I increase the signal intensity when I am using SuperSignal West Femto Maximum Sensitivity Substrate?

The following are possible causes for weak signal intensity:
- Low level of protein transferred from the gel to the membrane. This is particularly true for high molecular weight proteins. Reversible Protein Stain Kits for Nitrocellulose and PVDF membranes (Cat. Nos. 24580 and 24585, respectively) may be used to stain the bands on the membrane to indicate transfer efficiency.
- Over-diluted primary and secondary antibody probes. We suggest using the recommended dilution for the antibody probes for each substrate. More sensitive substrates will require a more dilute antibody solution.
- Short substrate incubation time. For best results, incubate the membrane for 5-15 min in the SuperSignal substrate.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How can I increase the signal intensity when I am using SuperSignal West Dura Extended Duration Substrate?

The following are possible causes for weak signal intensity:
- Low level of protein transferred from the gel to the membrane. This is particularly true for high molecular weight proteins. Reversible Protein Stain Kits for Nitrocellulose and PVDF membranes (Cat. Nos. 24580 and 24585, respectively) may be used to stain the bands on the membrane to indicate transfer efficiency.
- Over-diluted primary and secondary antibody probes. We suggest using the recommended dilution for the antibody probes for each substrate. More sensitive substrates will require a more dilute antibody solution.
- Short substrate incubation time. For best results, incubate the membrane for 5-15 min in the SuperSignal substrate.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Citations & References (1)

Citations & References
Abstract
A high-affinity reversible protein stain for Western blots.
Authors:Antharavally BS, Carter B, Bell PA, Krishna Mallia A
Journal:Anal Biochem
PubMed ID:15158487
'We describe a reversible staining technique, using MemCode, a reversible protein stain by which proteins can be visualized on nitrocellulose and polyvinylidine fluoride (PVDF) membranes without being permanently fixed to the membrane itself. This allows subsequent immunoblot analysis of the proteins to be performed. The procedure is applicable only to ... More