After staining my PVDF membrane with PageBlue Protein Staining Solution, I am getting high background on the membrane. Can you offer some tips?
This is most likely due to insufficient destaining time. We recommend destaining the membrane in 30% acetonitrile/20% ethanol solution for an additional 5 mins.