Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: This S5/1 monoclonal antibody recognizes human CD88 also known as C5aR1. C5aR1 is a complement receptor widely expressed on cells of the myeloid lineage. Clone S5/1 has been reported to block C5a binding.
Applications Reported: This S5/1 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This S5/1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
CD88, also known as C5aR1, is a C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. CD88 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, CD88 stimulates chemotaxis, granule enzyme release and superoxide anion production. CD88 is also a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with CD88 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5a-R; CD88 antigen; complement component 5a receptor 1
Gene Aliases: C5A; C5AR; C5R1; CD88
UniProt ID: (Human) P21730
Entrez Gene ID: (Human) 728
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support