L-Glutamine (200 mM), 100 mL - FAQs
View additional product information for L-Glutamine (200 mM) - FAQs (25030032, 25030024)
2 product FAQs found
What is your recommended method for thawing L-Glutamine before adding to cell culture media?
We recommend thawing at 2-8 degrees C and then warming at 37 degrees C until the material goes into solution. Mix it as it warms up to spend minimal time at elevated temperatures before you aliquot it.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Why did my 200 mM L-glutamine precipitate when I thawed it?
When L-glutamine is in a concentrated stock solution it easily precipitates when cooled. Warming the solution briefly in a 37 degrees C water bath with gentle swirling will dissolve the precipitate. Do not use the product unless the precipitate is fully dissolved.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.