Search
View additional product information for Nalgene™ PPCO Dilution Bottles with Closure - FAQs (2505-0380PK, 2505-0380)
1 product FAQs found
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.