Trypsin-EDTA(0.25%), phenol red
Gibco™

Trypsin-EDTA(0.25%), phenol red

This liquid formulation of trypsin contains EDTA and phenol red. Gibco Trypsin-EDTA is made from trypsin powder, an irradiated mixture자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
2520011420 x 100 mL
25200056100 mL
25200072500mL
카탈로그 번호 25200114
제품 가격(KRW)
373,000
온라인 행사
Ends: 31-Dec-2026
414,000
할인액 41,000 (10%)
Each
수량:
20 x 100 mL
Customize this product
제품 가격(KRW)
373,000
온라인 행사
Ends: 31-Dec-2026
414,000
할인액 41,000 (10%)
Each
This liquid formulation of trypsin contains EDTA and phenol red. Gibco Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements. See the complete range of Gibco trypsin solutions and recommended dissociation conditions.

This trypsin solution is modified as follows:
With
• EDTA
• Phenol Red

The complete formulation is available.

We offer a variety of trypsin formulations, including animal origin-free TrypLE reagents.

Quality testing
Gibco Trypsin solutions are tested for pH, osmolality, sterility, and performance. In addition, the raw materials are verified for e-beam irradiation and precursor lots are tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis.

Documented traceability
We can provide detailed documentation to meet your regulatory needs. Gibco Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.

cGMP manufacturing and quality system
For supply chain continuity, we manufacture Gibco Trypsin-EDTA (0.25%), phenol red at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
연구용 또는 추가 제조 공정용. 진단용으로는 사용할 수 없습니다.
사양
세포 유형포유류 세포
킬레이터EDTA
농도1X
삼투압270 ∼ 320 mOsm/kg
제품라인Gibco
수량20 x 100 mL
유통 기한24개월
소스동물 원료
테스트됨체외 바이오분석
분류동물 원료
형태액체
제품 유형세포 배양 해리 시약
멸균멸균 여과
Sterilization MethodSterile-filtered
첨가제 포함페놀 레드, EDTA, EDTA
pH7.1 ∼ 8
Unit SizeEach
구성 및 보관
보관 조건: -5°C ∼ -20°C
배송 조건: 얼음
유통 기한: 제조일로부터 24개월

자주 묻는 질문(FAQ)

Is Trypsin-EDTA (0.25%), phenol red (Cat. No. 25200114) made from the same trypsin powder as Trypsin (1:250), powder (Cat. No. 27250018)?

Yes. Trypsin-EDTA (0.25%), phenol red (Cat. No. 25200114) is made with the same trypsin powder as Trypsin (1:250), powder (Cat. No. 27250018).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep trypsin in the refrigerator after thawing?

Generally speaking, trypsin can be stored refrigerated for up to a week. However, this may vary depending on how often it is warmed up and used during that week. It may also depend on how tightly the cells are attached, because there will be some loss of activity each day the trypsin is not frozen.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between TrypLE Express Enzyme, TrypLE Select Enzyme, and CTS TrypLE Select Enzyme?

All the above products contain rProtease, a non-animal, trypsin-like enzyme formulation used for the dissociation of attachment-dependent cell lines. This alternative for porcine trypsin is a recombinant enzyme derived from microbial fermentation.

The TrpLE Express, TrypLE Select, and CTS TrypLE Select products differ in their production and testing methods and in the documentation provided.

TrypLE Select is for the industrial market and has release-testing both for endotoxins and enzyme activity. This material also has a Drug Master File on record. It is manufactured in AOF-dedicated equipment and is intended for Research Use/Further Cell Culture Manufacturing.

TrypLE Express is manufactured without use of dedicated equipment and is intended for Research Use Only. Not for use in diagnostic procedures.

CTS TrypLE Select is manufactured under scalable cGMP conditions using dedicated equipment and is designed for clinical applications. This product has a Drug Master File on record and is intended for Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. Not intended for direct administration into humans or animals.

To read more about TrypLE and cell release times using this reagent, please go here (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin/tryple-express.html#1).

Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.

What are the benefits of Gibco TrypLE reagent over trypsin?

TrypLE reagent is free of animal- and human-derived components, producing an exceptionally pure reagent that is gentle on cells. Inactivation with trypsin inhibitors is not required. TrypLE reagent is ideal for dissociating a variety of attachment-dependent mammalian cell lines both in serum and in serum-free conditions, and can be directly substituted for trypsin in your current protocol. TrypLE reagent is room-temperature stable and ready to use when you need it. TrypLE cell dissociation reagents remain stable for 24 months at room temperature, making storage and handling easier and more convenient.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
Self-renewal of embryonic-stem-cell-derived progenitors by organ-matched mesenchyme.
Authors:Sneddon JB, Borowiak M, Melton DA,
Journal:Nature
PubMed ID:23041930
One goal of regenerative medicine, to use stem cells to replace cells lost by injury or disease, depends on producing an excess of the relevant cell for study or transplantation. To this end, the stepwise differentiation of stem cells into specialized derivatives has been successful for some cell types, but ... More