Pierce™ Chromatin Prep Module
Thermo Scientific™

Pierce™ Chromatin Prep Module

The Thermo Scientific™ Pierce™ Chromatin Prep Module provides a simple, fast, and reproducible method to isolate chromatin for use inRead more
Have Questions?
Catalog NumberQuantity
2615830 reactions
Catalog number 26158
Price (USD)
423.00
Each
Quantity:
30 reactions
Request bulk or custom format
Price (USD)
423.00
Each
The Thermo Scientific™ Pierce™ Chromatin Prep Module provides a simple, fast, and reproducible method to isolate chromatin for use in assays that monitor transcription regulation through histone modification1 (epigenetics) or transcription factor-DNA binding interactions, such as chromatin immunoprecipitation (ChIP). The module contains all reagents necessary to isolate crosslinked chromatin from mammalian cells.

This Chromatin Prep Module is also included as part of our complete Agarose ChIP Kit.

Related Products
Pierce™ Agarose ChIP Kit
Pierce™ ChIP-Grade Protein A/G Plus Agarose
Proteinase K
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Chromatin Prep Module
FormatKit
Quantity30 reactions
Sufficient For30 Reactions
Binding Capacity2 x 106 Cells/Reaction
Product LinePierce
TypeChromatin Isolation Kit
Unit SizeEach
Contents & Storage
Sufficient For: 30 chromatin preparations for ChIP assays
• Membrane Extraction Buffer, 10 mL
• Nuclear Extraction Buffer, 10 mL
• MNase Digestion Buffer, 5 mL
• MNase Stop Solution, 0.5 mL
• Halt Protease and Phosphatase Inhibitor Cocktail EDTA-free (100X), 4 x 90 μL
• Sodium chloride (5M), 3 mL
• Glycine solution (10X), 15 mL
• PBS (20X), 15 mL
• Micrococcal Nuclease (ChIP Grade) (10 U/μL) (store at -20°C)
• Proteinase K (20 mg/mL), 0.25 mL (store at -20°C)
• DTT, Lyophilized, 1 vial

Unless otherwise indicated, store all components at 4°C.