Search
View additional product information for Pierce™ HA-Tag IP/Co-IP Kit - FAQs (26180)
1 product FAQs found
The HA-tagged Positive Control is an E. coli lysate with a total protein concentration of 1 mg/mL containing HA-tagged GSTPI3K-SH2 domain fusion protein of around 37 kDa.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.