Pierce™ HA-Tag IP/Co-IP Kit
Thermo Scientific™

Pierce™ HA-Tag IP/Co-IP Kit

The Thermo Scientific Pierce HA Tag IP/Co-IP Kit provides the affinity resin, positive control and other reagents necessary to performRead more
Have Questions?
Catalog NumberQuantity
2618025 reactions kit
Catalog number 26180
Price (USD)
730.00
Each
Quantity:
25 reactions kit
Request bulk or custom format
Price (USD)
730.00
Each
The Thermo Scientific Pierce HA Tag IP/Co-IP Kit provides the affinity resin, positive control and other reagents necessary to perform immunoprecipitation (IP) or co-immunoprecipitation (Co-IP) reactions with an HA-tagged bait protein.

Features of the HA-Tag IP/Co-IP Kit:

• Improved—updated kit includes more immobilized antibody resin and improved protocol along with GST-PI3K-SH2-HA fusion protein as a positive control
• Specific—highly specific anti-HA monoclonal antibody provides high-yield immunoprecipitation products and clean Western blot detection
• Compatible—includes protocols and reagents for multiple elution conditions to accommodate different protein sensitivities and downstream applications
• Robust—the kit and the anti-HA agarose are compatible with various cell lysates and physiologic (non-denaturing) buffer systems
• Convenient and easy—complete kit includes all necessary reagents, convenient spin columns and easy-to-follow instructions

This kit is based on crosslinked beaded agarose to which a highly specific anti-HA antibody is covalently immobilized. The ready-to-use affinity resin, together with the included buffers, microcentrifuge spin columns, positive control, and easy-to-follow instructions, constitute a complete set of reagents sufficient for 25 for IP or Co-IP assays. Upon incubation with a sample containing the tagged fusion protein, interaction complexes involving the HA-tagged bait protein are captured on the agarose beads. After simple washing steps, the specific protein interaction complex is easily eluted from the resin in the supplied elution buffer or SDS-PAGE sample loading buffer for subsequent analysis.

The hemagglutinin (HA) peptide (YPYDVPDYA), derived from the human influenza virus HA protein is one of several fusion protein tags used for recombinant protein expression. Utilizing a specific, high-affinity immobilized antibody, HA-tagged fusion proteins can be quickly purified from bacterial and mammalian cell lysates as well as from the Pierce Human in vitro translation reactions. For co-immunoprecipitation reactions, simple wash steps allow enrichment and elution of specific protein interaction complexes into the supplied elution buffer or SDS-PAGE sample loading buffer for subsequent analysis.

Related Products
Pierce™ HA-Tag Magnetic IP/Co-IP Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayAffinity Purification
For Use With (Application)Pull-Down (Tag-Based), Immunoprecipitation (IP)
For Use With (Equipment)Microcentrifuge
FormatKit
Quantity25 reactions kit
Sufficient For25 Reactions
TargetTarget-Specific
Product LinePierce
TypeTag IP/Co-IP Kit
Unit SizeEach
Contents & Storage
Sufficient For: 25 IP/Co-IP reactions, each using 10 μL of resin
• Anti-HA Agarose, 0.25 mL
• HA-tagged Positive Control, 0.5 mL
• BupH Tris Buffered Saline Pack (500 mL), 1 pack
• Lane Marker Non-Reducing Sample Buffer (5X), 5 mL
• Pierce Spin Columns Accessory Pack, 27 columns
• Collection Tubes and Caps Accessory Pack, 100 tubes

Store HA-tagged positive control at -80°C (may be stored at -20°C for up to 6 months). Store remaining kit components at 4°C.

Frequently asked questions (FAQs)

What is the molecular weight of the HA-Tagged Positive Control within the Pierce HA-Tag IP/Co-IP Kit (Cat. No. 26180)?

The HA-tagged Positive Control is an E. coli lysate with a total protein concentration of 1 mg/mL containing HA-tagged GSTPI3K-SH2 domain fusion protein of around 37 kDa.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.