Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IRELEDKKK IQNLLALVGT DAGEVTYFCK EPPHKVTILQ KTIQAVGECE QSESSAFKAD PKISKRRPSR ERKESSEHYQ RDIQTLILQV EALQAQLGEQ TKLSREQIEG LIEDRRIHLE EIQVQHQRNQ NKIKELTKNL HHTQELLYES TKDFLQLRSE NQNKEKSWML EKDNLMSKIK QYRVQCKKKE DKIGKVLPVM HESHHAQSEY IKSLKDKLVQ EKKLSNMYQE QCISLEEELA RIREEEGMRR EIFKDRTNKM GKRLQIMTKR YEALERRRIL EVEGFKTDIK VLRQKLKDLE QMLYKATVNA RANQDLALLC EVRDSNRRAH KIQGELKNLK SKVFGLENEL RLC (137-488 aa encoded by BC006444)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 77; unnamed protein product
Gene Aliases: CCDC77
UniProt ID: (Human) Q9BR77
Entrez Gene ID: (Human) 84318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support