Search
View additional product information for 96 Well White/Clear Bottom Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10 - FAQs (265302)
1 product FAQs found
Opaque plates can only be used on top reader instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.